Filtered Search Results
Methanol, UHPLC-MS
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Unique signal-to-noise specification for the chemical industry where the solvent quality is linked directly to the sensitivity of the mass spectrometer. | CAS: 67-56-1 | CH4O | 32.04 g/mol
| PubChem CID | 887 |
|---|---|
| CAS | 67-56-1 |
| Molecular Weight (g/mol) | 32.04 |
| ChEBI | CHEBI:17790 |
| MDL Number | MFCD00004595 |
| SMILES | CO |
| Synonym | methyl alcohol,wood alcohol,carbinol,wood spirit,wood naphtha,methylol,methyl hydroxide,pyroxylic spirit,colonial spirit,columbian spirit |
| IUPAC Name | methanol |
| InChI Key | OKKJLVBELUTLKV-UHFFFAOYSA-N |
| Molecular Formula | CH4O |
Acetonitrile, UHPLC-MS
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Unique signal-to-noise specification for the chemical industry where the solvent quality is linked directly to the sensitivity of the mass spectrometer. | CAS: 75-05-8 | C2H3N | 41.053 g/mol
| PubChem CID | 6342 |
|---|---|
| CAS | 75-05-8 |
| Molecular Weight (g/mol) | 41.053 |
| ChEBI | CHEBI:38472 |
| SMILES | CC#N |
| Synonym | methyl cyanide,cyanomethane,ethanenitrile,methanecarbonitrile,ethyl nitrile,methane, cyano,acetonitrile,cyanure de methyl,methylkyanid,mecn |
| IUPAC Name | acetonitrile |
| InChI Key | WEVYAHXRMPXWCK-UHFFFAOYSA-N |
| Molecular Formula | C2H3N |
Water, UHPLC-MS
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Unique signal-to-noise specification for the chemical industry where the solvent quality is linked directly to the sensitivity of the mass spectrometer. | CAS: 7732-18-5 | H2O | 18.015 g/mol
| PubChem CID | 962 |
|---|---|
| CAS | 7732-18-5 |
| Molecular Weight (g/mol) | 18.015 |
| ChEBI | CHEBI:15377 |
| SMILES | O |
| Synonym | dihydrogen oxide,dihydrogen monoxide |
| IUPAC Name | oxidane |
| InChI Key | XLYOFNOQVPJJNP-UHFFFAOYSA-N |
| Molecular Formula | H2O |
ChromaCare™ LC-MS Instrument Flush Solution
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
ChromaCare™ LC-MS Instrument Flush Solution is designed to prepare LC/MS instruments for start-up. By reducing background noise, this solution facilitates instrument installation and preventative maintenance routines. ∣ CAS: 75-05-8; 67-56-1; 67-63-0; 7732-18-5
| Percent Purity | 25% Acetonitrile, 25% Methanol, 25% Water, 25% 2-Propanol (IPA) |
|---|---|
| CAS | 75-05-8; 67-56-1; 67-63-0; 7732-18-5 |
| Color | Colorless |
| Physical Form | Liquid |
| Grade | LC-MS |
Thermo Scientific™ UHPLC-MS Reagents Installation Kit
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
ChromaCare™ LC-MS Aqueous Rinse, Probe Wash 1, Thermo Scientific™
Ready-to-use blend of water and 2% acetonitrile (v/v) serves as an aqueous wash solution to clean sample probes and needles on LC instruments.
Ammonium Formate in Methanol, 10mM, with 0.05% Formic Acid, LC-MS
Mobile phase blend suitable for analysis such as vitamin D and drugs of abuse testing, and packaged in a borosilicate glass bottle that can be used directly on the LC-MS instrument. | CAS: 540-69-2 | CH4O | 32.04 g/mol
| Name Note | Mobile-phase solvent blend |
|---|---|
| CAS | 64-18-6 |
| Chemical Name or Material | Ammonium Formate in Methanol, 10mM, with 0.05% Formic Acid |
Ammonium Formate in Water, 10mM, with 0.05% Formic Acid, LC-MS
Mobile phase blend suitable for analysis such as vitamin D and drugs of abuse testing, and packaged in a borosilicate glass bottle that can be used directly on the LC-MS instrument. | CAS: 7732-18-5
| Name Note | Mobile-phase solvent blend |
|---|---|
| CAS | 64-18-6 |
| Chemical Name or Material | Ammonium Formate in Water, 10mM, with 0.05% Formic Acid |
ChromaCare™ LC-MS Biologics Flush Solution
Flush solution for cleaning blood, plasma, and urine samples from the chromatograph.
| Percent Purity | Pass Test (By GC-FID) |
|---|---|
| CAS | 75-05-8 |
| Physical Form | Liquid |
| pH | 6 to 7 |
| Grade | LC-MS |
| Synonym | isopropanol,2-propanol,isopropyl alcohol,2-hydroxypropane,alkolave,avantine,hartosol,dimethylcarbinol,sec-propyl alcohol,petrohol |
ChromaCare™ LC-MS Organic Rinse, Probe Wash 2, Thermo Scientific™
Premade and ready-to-use water-acetonitrile blend (60:40) with 0.1% formic acid serves as a wash solution to clean sample and liquid chromatography injection probes. Commonly used as an auto-sampler cleaning solution in LC-MS.