Log into Your Account
Don't have a profile? Register
Forgot Password?
Search
Customer Service:  800-766-7000Web Support:  877-885-2081All Other Contact Info
Refine Browse Results
Browse Results: 11 item(s)
You're Browsing:QC/QAP
  1. 1
1 Fisher Diagnostics* HDL Controls
HDL Control High HDL Control Medium HDL Control Low
2 Fisher Diagnostics* HDL-C/LDL-C Calibrator
HDL-C/LDL-C Calibrator
3 Fisher Diagnostics* Lipid Control
Levels 1 and 2; 5 x 3mL each level
4 Thermo Scientific* MAS* Cerebrospinal Fluid Controls
Liquid assayed cerebrospinal fluid controls   ·  Liquid, ready-to-use control eliminates variables due to reconstitution, mixing errors, or uncertain water quality   ·  Contains 18 analytes including Glutamine and Potassium
5 MAS* Lyo IAC
A lyophilized multi-analyte control to help monitor immunoassay and therapeutic drug monitoring methods   ·  Liquid, ready to use; no preparation required   ·  Human serum based   ·  3 significant levels available Level 2 Level 1 Level 3
6 Thermo Scientific* MAS* Diabetes Controls
Bi-level control designed to monitor hemoglobin A1c methods   ·  Liquid, ready-to-use  ·  Monitor laboratory testing procedures for Hemoglobin A1c  ·  Human whole blood matrix  ·  Assayed values available for most major analyzers  ·  
7 MAS* CardioImmune*-XAB Cardiac Marker Control
Multi-analyte control   ·  Stable liquid control   ·  CardioImmune-XAB assayed for the Abbott AxSYM*   ·  Human serum based   ·  30-day open-vial stability for all analytes at 2°–8°C   ·  90-day unopened-vial stability for all analytes at 2°–8°C
8 MAS* CardioImmune* TL Cardiac Marker Control
Assayed control serum suitable for monitoring assay conditions in specific cardiac marker determinations   ·  Stable liquid control   ·  Human serum based   ·  Assayed for major automated platforms   ·  3-year shelf life at -20°C   ·  180-day unopened-vial stability
9 MAS Dropper Tips
Dropper Tips
10 MAS* Lipid Control
Bi-level, assayed control offering a complete lipid profile Assayed ranges for precipitate and direct HDL/LDL methods Liquid, ready-to-use control eliminates variables due to reconstitution, mixing errors, or uncertain water quality Level 2; 6 x 3mL Level 1; 6 x 3mL
11 Thermo Scientific* MAS* Liquimmune* Immunoassay Control
Multi-analyte control to help monitor immunoassay methods   ·  Liquid, ready-to-use  ·  Human serum-based  ·  Suitable for monitoring assay conditions in many immunochemistry and therapeutic drug determinations  ·  Assayed values available
  1. 1