Refine Browse Results
Browse Results: 754 item(s)
You're Browsing: Chemicals
Quality chemistry
We manufacture our own chemical brands, such as FisherChemical and Acros Organics, to the highest quality standards in FDA-licensed, ISO-certified, cGMP facilities. Validated chemical solutions specific for the food industry such as Babcock testing, Mojonnier testing and Karl Fischer reagents are standards in the FisherChemical portfolio.
1
Arsenic, Reference Standard Solution (1000ppm ±1%/Certified), Fisher Chemical
7697-37-2 1327-53-3 7732-18-5 For atomic absorption. (1mL = 1mg As) Solute: Arsenic Trioxide Solvent: Dilute Nitric Acid.
2
Barium Acetate (Crystalline/Certified ACS), Fisher Chemical
543-80-6 C4H6O4Ba Ba(C2 H3O2 )2 Acetic Acid Barium Salt
3
Calcium Lactate Pentahydrate (USP), Fisher Chemical
5743-47-5 C6H10O6Ca·5H2 O Ca(CH3CH(OH)CO2 )2 ·5H2 O Lactic Acid Calcium Salt
4
Arsenic Trioxide (Primary Standard), Fisher Chemical
1327-53-3 As2O3 Arsenous Acid
5
Vanadium, Reference Standard Solution (1000ppm ±1%/Certified), Fisher Chemical
7647-01-0 1314-62-1 7732-18-5 For atomic absorption.
6
Hydrochloric Acid Concentrate, (Certified), Fisher Chemical
7647-01-0 7732-18-5 HCl Muriatic Acid Each bottle contains exact amount of 1N concentrate required to make 1 liter of 0.1N (N/10) solution.
7
Manganese (II) Sulfate Solution (APHA), Fisher Chemical
7785-87-7 7732-18-5 For determination of dissolved oxygen.
8
Lactic Acid (USP/FCC), Fisher Chemical
50-21-5 97-73-4 C3H6O3 CH3CH(OH)CO2 H 2-Hydroxypropanoic Acid
9
Calcium Gluconate Monohydrate (Powder/USP), Fisher Chemical
18016-24-5 C12 H2 2 O14Ca·H2 O Glucobiogen D-Gluconic Acid, Calcium Salt Glucal
10
Potassium Hydroxide Solution, 45% w/w (Certified), Fisher Chemical
1310-58-3 7732-18-5 KOH
11
Mercuric Acetate (Crystalline/Certified ACS), Fisher Chemical
1600-27-7 C4H6HgO4 (CH3COO)2 Hg Acetic Acid Mercury Salt Mercury (II) Acetate
12
Ethyl Acetate (HPLC), Fisher Chemical
141-78-6 C4H8O2 CH3COOC2 H5 EtOAc Acetic Acid Ethyl Ester HPLC mobile phase for analytical and preparative separations.
13
Buffer Solution, pH 7.00 (Certified), Fisher Chemical
7778-77-0 1310-73-2 7732-18-5 Chlorine Titrimeter Reagent Potassium Phosphate Monobasic/Sodium Hydroxide.
14
Ammonium Chloride (Granular/USP/FCC), Fisher Chemical
12125-02-9 NH4Cl
15
Buffer Concentrate, pH 10.00 (Certified), Fisher Chemical
7447-40-7 1310-73-2 139-33-3 7732-18-5 10043-35-3 Boric Acid/Potassium Chloride/Sodium Hydroxide.