Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Refine Browse Results
Browse Results: 15 item(s)
You're Browsing:Forceps and Scalpels
  From general-purpose forceps and scalpels to surgical-grade instruments for specialized applications, we have the products you need to support successful tissue handling and cell culture.
  1. 1
1 Available on GSA/VA Contract for Federal Government customers only. Fisherbrand* Stainless-Steel Blades for Dissecting Knife Handles
Exceptionally strong and uniformly sharp   ·  Surgical quality  ·  Sterile  ·  Individually foil-wrapped  ·   Blade size: 21; Length: 1.562 in. (4cm) Blade size: 15; Length: 1.375 in. (3.5cm) Blade size: 22; Length: 1.375 in. (3.5cm) Blade
2 Available on GSA/VA Contract for Federal Government customers only. Fisherbrand* Single-Use Scalpels
Surgical quality—also ideal for student or general lab use   ·  Stainless-steel blade attached to plastic handle  ·  Sterile, individually packaged  ·  Plastic guard protects cutting edge and prevents package puncture  ·   Includes
3 Available on GSA/VA Contract for Federal Government customers only. Fisherbrand* General-Purpose Lab Knife Set
For slicing rubber tubing, corks, plant samples, and paraffin Blades   ·  Made of the highest-grade steel  ·  Ultramicro to heavy-duty cutting edges  ·   Handles   ·  Eight
4 Fisherbrand* Dissecting Dressing Forceps
Treated for increased resistance to corrosion   ·  Can be sterilized by EtO/gamma radiation  ·  Rockwell hardness (HRC): 40–45  ·  Electronically passivated  ·  Autoclavable up to 135°C  ·  Latex-free  ·  Forged,
5 Fisherbrand* Disposable-Blade Dissecting Knife Handles
Smooth, tapered back end serves as blunt probe Reusable handles in a choice of styles sold separately from interchangeable blades (see Fisherbrand Stainless-steel Blades). Stainless-steel handles scientifically shaped and balanced for delicate dissecting and surgical use.   ·  Blades
6 Fisherbrand* Dissecting Self-Closing Forceps
Sharp-pointed tips are normally closed   ·  Can be sterilized by EtO/gamma radiation  ·  Rockwell Hardness (HRC): 45  ·  Passivated  ·  Autoclavable  ·  Latex-free  ·  Stainless steel per ASTM* standard  ·  Squeeze
7 Fisherbrand* Dissecting Tissue Forceps
ASTM* standard stainless steel   ·  Can be sterilized by EtO/gamma radiation  ·  Rockwell hardness (HRC): 40–45  ·  Passivated  ·  Autoclavable up to 135°C  ·  Latex-free  ·  1 x 2 teeth grip tissue
8 Fisherbrand* Dissecting Jewelers Microforceps
For very precise work   ·  Can be sterilized by EtO/gamma radiation  ·  Rockwell hardness (HRC): 40–45  ·  Passivated  ·  Fully autoclavable  ·  Latex-free  ·  ASTM* standard stainless steel  ·  Ultrathin,
9 Fisherbrand* Dissecting Fine-Pointed Forceps
Serrated tips with guide pins   ·  Can be sterilized by EtO/gamma radiation  ·  Rockwell hardness (HRC): 40–45  ·  Passivated  ·  Fully autoclavable  ·  Latex-free  ·  Patterned grip handles  ·  
10 Fisherbrand* Dissecting Blunt-Pointed Forceps
Heavy steel construction   ·  ASTM* standard stainless steel  ·  Can be sterilized by EtO/gamma radiation  ·  Rockwell hardness (HRC): 40–45  ·  Electronically passivated  ·  Autoclavable  ·  Latex-free  ·  Highly
11 Fisherbrand* Curved, Serrated or Smooth Tip Microforceps
ASTM* (F899-84) standard stainless steel   ·  Can be sterilized by EtO/Gamma radiation  ·  Rockwell hardness (HRC): 40–45  ·  Passivated  ·  Fully autoclavable  ·  Patterned grip handles  ·   Length:
12 Fisherbrand* Medium-Pointed Forceps
ASTM* standard stainless-steel   ·  Can be sterilized by EtO/gamma radiation  ·  Rockwell hardness (HRC): 40–45  ·  Passivated  ·  Fully autoclavable  ·  Latex-free  ·   Straight; Length: 4.5 in.
13 Fisherbrand* Dissecting Micro-Adson Serrated Tips Forceps
ASTM* standard stainless steel   ·  Can be sterilized by EtO/gamma radiation  ·  Rockwell hardness (HRC): 40–50  ·  Passivated  ·  Fully autoclavable  ·  Latex-free  ·  Delicate—very easy to press
14 Fisherbrand* Dissecting Extra-Fine-Pointed Splinter Forceps
Treated for increased resistance to corrosion   ·  Can be sterilized by EtO/gamma radiation  ·  Rockwell hardness: 40–45 HRC  ·  Passivated  ·  Fully autoclavable  ·  Surgical-quality stainless steel  ·  Straight,
15 Fisherbrand* Dissecting Cushing Delicate Thumb Forceps
Stainless-steel construction per ASTM* standard   ·  Can be sterilized by EtO/gamma radiation  ·  Rockwell hardness (HRC): 40–45  ·  Passivated  ·  Fully autoclavable  ·  Latex-free  ·  Patterned grip
  1. 1