Log into Your Account
Don't have a profile? Register
Forgot Password?
Search
Customer Service:  800-766-7000Web Support:  877-885-2081All Other Contact Info
Browse Results: 282 item(s)
You're Browsing:Remel Prepared Microbiology Media
  1. 1
  2. 2
  3. 3
  4. 4
  5. 5
  6. >
  7. >>
1 Thermo Scientific* Remel* Amies Transport Medium w/o Charcoal
Amies Transport Medium w/o Charcoal
2 Thermo Scientific* Remel* Potato Dextrose Agar (pH 3.5)
(pH 3.5) For cultivation, identification, and enumeration of yeast and fungi Potato Dextrose Agar pH 3.5.
3 Thermo Scientific* Remel* Motility Test Medium w/TTC Indicator
Motility Test Medium w/TTC Motility Test Medium w/TTC
4 Thermo Scientific* Remel* Bacteroides Fragilis Isolation Agar (BBE Agar)
(BBE Agar) For isolation and presumptive identification of Bacteroides fragilis group Bacteroides Fragilis Isolation Agar (BBE Agar); Monoplate; 10/Pk
5 Thermo Scientific* Remel* Strep Selective Agar (w/RNA, maltose, neomycin, polymyxin B)
w/RNA, maltose, neomycin, polymyxin B. For primary isolation of group A beta hemolytic streptococci from upper respiratory specimens   ·  Neomycin and polymyxin B suppress much of the oral flora, including coliforms, staphylococci, Micrococcus, Haemophilus,
6 Thermo Scientific Remel* Phenylalanine Agar
Phenylalanine Agar Phenylalanine Agar; 20/Pack
7 Thermo Scientific* Remel* Anaerobic LKV Blood Agar (CDC formulation)
CDC formulation; Laked blood enhances pigmentation of anaerobic microorganisms   ·  Kanamycin and vancomycin inhibit gram-positive microorganisms and facultative anaerobic bacteria, and select for gram-negative bacilli  ·   Anaerobic LKV Blood Agar; Format: Monoplate;
8 Thermo Scientific* Remel* Brucella Broth w/.16% Agar, 1% Glycine
Brucella Broth w/0.16% Agar; 1%Glycine
9 Thermo Scientific Remel*Anaerobic KV Blood Agar (CDC formulation)
CDC formulation; Kanamycin and vancomycin inhibit gram-positive microorganisms and facultative anaerobic bacteria, and select for gram-negative bacilli Anaerobic KV Blood Agar; Format: Monoplate; pack of 100 Anaerobic KV Blood Agar; Monoplate; 10/Pack
10 Thermo Scientific* Remel* Strep Selective II Agar (w/colistin, oxolinic acid)
w/colistin, oxolinic acid. For primary isolation of group A beta hemolytic streptococci of medical and veterinary importance   ·  Colistin and oxolinic acid allow growth of streptococci while inhibiting staphylococci, coryneforms, and gram-negative microorganisms  ·  
11 Thermo Scientific* Remel* Anaerobic Blood Agar (CDC formulation)
CDC formulation; For primary cultivation and isolation of fastidious anaerobic microorganisms Anaerobic Blood Agar; Monoplate; 100/Case Anaerobic Blood Agar; Spacesaver Plate; 15/Pack Anaerobic Blood Agar; Monoplate; 10/Pack
12 Thermo Scientific* Remel* CTA w/1% Arabinose (Cystine Trypticase Agar)
For determination of fermentation reactions by fastidious microorganisms such as Neisseria, pneumococci, streptococci and nonsporeforming anaerobes CTA w/1% Arabinose (Cystine Trypticase Agar)
13 Thermo Scientific* Remel* TGEA
(Tryptone Glucose Extract Agar) For cultivation and enumeration of bacteria in water, milk and other dairy products. Monoplate; 100/Pk. Monoplate; 10/Pk.
14 Thermo Scientific Remel* Casamino Acid Solution (1%)
Casamino Acid Solution 1%
15 Thermo Scientific Remel*PRAS Indole Nitrate Medium (VPI) (Pre-Reduced Anaerobic Sterilized)
P.R.A.S. Indole Nitrate Medium ; Format: 16 x 125mm tube; Volume: 5mL; pack of 20
  1. 1
  2. 2
  3. 3
  4. 4
  5. 5
  6. >
  7. >>