Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Browse Results: 39 item(s)
You're Browsing:Antibody Production and Purification
  1. 1
  2. 2
  3. 3
  4. >
1 Thermo Scientific* Pierce* IgG Plus Orientation Kits
Crosslink oriented antibodies to agarose resin to create antigen affinity column These IgG orientation kits use a crosslinker to covalently attach antibody that is bound to Protein A or Protein G agarose resin, thereby creating a reusable affinity column for purification of antigens. The Plus Orientation
2 Thermo Scientific* Pierce* Imject* Maleimide-Activated mcKLH and Kit
Cysteine-reactive KLH for immunogen preparation with sulfhydryl peptides The high-quality mariculture KLH is rendered sulfhydryl-reactive by chemical modification with Sulfo-SMCC crosslinker, and the resulting maleimide-activated protein is then stabilized and lyophilized in phosphate buffer. Maleimide
3 Thermo Scientific* Pierce* High-Capacity Protein A Agarose and Kits
Resin and columns for high-yield human or rabbit IgG purification at any scale Pierce Protein A Plus Agarose consists of purified native Protein A that has been covalently immobilized at high density onto high-quality crosslinked 6% beaded agarose (CL-6B). Among the many available forms of immobilized
4 Thermo Scientific* Pierce* Binding and Elution Buffers for IgG Purification
Ensure optimal binding and elution conditions for affinity applications These products include IgG Binding Buffers specific for each immunoglobulin-binding protein (Protein A, Protein G, Protein A/G). The Pierce IgG Elution Buffer provides efficient dissociation and elution of antibodies from all three
5 Thermo Scientific* Pierce* Imject* Mariculture KLH and EDC Conjugation Kit
The most immunogenic carrier protein for antigen prep and antibody production Mariculture KLH (mcKLH) is purified and lyophilized in buffers that optimize its stability and solubility for hapten conjugation. The large, complex protein contains hundreds of sites per molecule for effective conjugation
6 Thermo Scientific* Pierce Melon Gel* Monoclonal IgG Purification Kit
High-yield, mild purification system for mouse monoclonal antibodies The Melon Gel Monoclonal IgG Purification Kit consists of a large package size of Melon Gel Resin and an optimized protocol for easy and mild purification of monoclonal IgG antibodies from hybridoma culture supernatant or ascites fluid.
7 Thermo Scientific* SulfoLink* Coupling Resins and Immobilization Kits
Immobilize sulfhydryl-containing peptides or proteins for affinity purification SulfoLink Coupling Resin is porous, crosslinked 6% beaded agarose that has been activated with iodoacetyl groups. When incubated with a solution of peptide or protein that contains reduced cysteine residues, the iodoacetyl
8 Thermo Scientific* Nunc* ProPur Antibody Purification Spin Columns
Designed for simple, rapid antibody purification from serum, ascites, and tissue culture supernatant Convenient: No previous chromatography experience required. Rapid: Complete purification and concentration in 50
9 Thermo Scientific* Pierce* Imject* BSA Protein and EDC Conjugation Kit
Popular carrier protein for conjugation to haptens and other weak antigens Imject BSA is purified and lyophilized in buffers that optimize its stability and solubility for hapten conjugation. The large, single polypeptide protein contains numerous sites per molecule for effective conjugation of peptides
10 Thermo Scientific* Pierce* IgM Purification Kit
Easy-to-use affinity matrix and buffer system to purify IgM Pierce MBP Agarose is purified mannan binding protein (also called mannose-binding lectin, MBL) that has been immobilized on 4% beaded agarose. When used with an optimized buffer system based on temperature- and calcium-dependent binding, this
11 Thermo Scientific* Pierce* Protein G Agarose and Kits
Resin and columns for human and mouse IgG purification at any scale Pierce Protein G Agarose is effective for affinity purification of IgG from serum and other fluids of many mammalian species. Protein G is especially well suited for use with mouse antibodies (including IgG1) in addition to most IgG
12 Thermo Scientific* Pierce* Thiophilic Adsorbent Resin and Kit
A non-protein immunosorbent for the purification of immunoglobulins Pierce Thiophilic Adsorbent is 6% beaded agarose that has been modified to contain simple sulfone-thioether groups. The result is an affinity resin that has a high binding capacity and a broad specificity for many species of immunoglobulins
13 Thermo Scientific* Pierce* Protein A/G Agarose and Kits
Resin and columns for broad-spectrum IgG purification at any scale Pierce Protein A/G Agarose binds nearly all isotypes and mammalian species of IgG from serum, ascites fluid, cell culture supernantant and other antibody samples, thereby enabling effective antibody purification. Because the immobilized
14 Thermo Scientific* Protein L Resins and Kits
Resin and columns for immunoglobulin purification at any scale Pierce Protein L Agarose is a versatile affinity resin for purification of mammalian IgG isotypes that contain specific kappa light chains. Protein L is especially suited for isolating mouse IgG VkI and human IgG VkI, VkIII and VkIV monoclonal
15 Thermo Scientific* Pierce* Imject* Maleimide-Activated BSA and Kit
Cysteine-reactive BSA for immunogen preparation with sulfhydryl peptides Purified, carrier-grade BSA is rendered sulfhydryl-reactive by chemical modification with Sulfo-SMCC crosslinker, and the resulting maleimide-activated protein is then stabilized and lyophilized in phosphate buffer. Maleimide groups
  1. 1
  2. 2
  3. 3
  4. >