Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Refine Browse Results
Browse Results: 46 item(s)
You're Browsing:Protein Purification
  1. 1
  2. 2
  3. 3
  4. 4
  5. >
1 Thermo Scientific* Pierce* Avidin Agarose
Strong biotin interaction creates a nearly irreversible bond This avidin affinity resin is crosslinked beaded agarose onto which purified avidin has been covalently immobilized. The biotin-binding agarose beads are available as pre-packed columns or resin slurry. The resin is designed for use in a variety
2 Thermo Scientific* Pierce* High-Capacity Protein A Agarose and Kits
Resin and columns for high-yield human or rabbit IgG purification at any scale Pierce Protein A Plus Agarose consists of purified native Protein A that has been covalently immobilized at high density onto high-quality crosslinked 6% beaded agarose (CL-6B). Among the many available forms of immobilized
3 Available on GSA/VA Contract for Federal Government customers only. 5 PRIME* PerfectPro* Ni-NTA Superflow
Ni-NTA resin with high binding capacity for efficient purification of 6xHis-tagged proteins by FPLC PerfectPro Superflow is a nickel-charged affinity resin that can be used to purify recombinant proteins containing a polyhistidine (6xHis) sequence. PerfectPro Superflow combines superior mechanical stability
4 Thermo Scientific* Pierce* High-Capacity Protein G Agarose
High-density immobilized Protein G beaded agarose resin Pierce Protein G Plus Agarose consists of recombinant Protein G that has been covalently immobilized at high density onto high-quality crosslinked 6% beaded agarose (CL-6B). The resin is effective for affinity purification of IgG from serum and
5 Thermo Scientific* SulfoLink* Coupling Resins and Immobilization Kits
Immobilize sulfhydryl-containing peptides or proteins for affinity purification SulfoLink Coupling Resin is porous, crosslinked 6% beaded agarose that has been activated with iodoacetyl groups. When incubated with a solution of peptide or protein that contains reduced cysteine residues, the iodoacetyl
6 Thermo Scientific* Pierce* c-Myc Tag IP/Co-IP Kit
Uses high-affinity antibodies to efficiently isolate c-Myc-tagged proteins and interactors The c-Myc peptide (EQKLISEEDL) has become a popular fusion tag for mammalian recombinant protein expression, and this kit facilitates immunoprecipitation (IP) or co-immunoprecipitation (co-IP) experiments involving
7 Thermo Scientific* Pierce* Resin Separators
Decant solutions after resin incubations performed in test tubes Resin Separators are ideal for separating beaded agarose and other affinity resins from liquid samples in methods that have been performed in 16mm x 100mm test tubes. Simply slide the resin separator into a test tube containing the support
8 Thermo Scientific* Pierce* NeutrAvidin* Resins
The high-performance, lower-cost alternative to streptavidin resins NeutrAvidin Agarose and UltraLink* Resins are superior alternatives to avidin and streptavidin beads in many biotin-based affinity applications. NeutrAvidin Protein is a modified avidin derivative that combines several key
9 Thermo Scientific* Pierce* High-Capacity Protein G UltraLink* Resin
A durable polyacrylamide resin for Protein G chromatography The Protein G Plus UltraLink Resin is used for high-capacity binding and purification of IgG in gravity-flow, spin-column or medium-pressure chromatography methods. Protein G is a popular antibody-binding protein that is especially useful for
10 Available on GSA/VA Contract for Federal Government customers only. 5 PRIME* PerfectPro* Ni-NTA MagBeads
Magnetic Ni-NTA resin for high-throughput, microscale purification of 6xHis-tagged proteins PerfectPro Ni-NTA MagBeads can be used for capturing 6xHis-tagged proteins under either native or denaturing conditions. PerfectPro Ni-NTA MagBeads are supplied as a 5% (v/v) suspension with a binding capacity
11 Thermo Scientific* Pierce* ConA and WGA Glycoprotein Isolation Kits
Isolate glycoproteins from complex protein mixtures in less than an hour These two kits use lectins concanavilin A (ConA) and wheat germ agglutinin (WGA), respectively, as ligands to bind specific classes of glycoproteins from complex protein samples. ConA lectin recognizes alpha-linked mannose and
12 Thermo Scientific* Pierce* D-Galactose Agarose
Purify lectins, galactosidases and other galactose-binding molecules Immobilized D-Galactose is a beaded agarose affinity resin that enables purification of agglutinins, lectins, toxins, and other galactose-binding proteins, N-acetylgalactosamine-binding proteins or carbohydrate-binding
13 Thermo Scientific* Pierce* Protein G Agarose and Kits
Resin and columns for human and mouse IgG purification at any scale Pierce Protein G Agarose is effective for affinity purification of IgG from serum and other fluids of many mammalian species. Protein G is especially well suited for use with mouse antibodies (including IgG1) in addition to most IgG
14 Thermo Scientific* Pierce* Streptavidin Resins
Resins with high biotin affinity and low nonspecific binding Pierce Streptavidin Agarose and UltraLink Resins supply immobilized streptavidin biotin-binding protein in different resin formats, binding capacities and package sizes to suit nearly any immunoprecipitation or affinity purification procedure
15 Thermo Scientific* Pierce* GST Agarose
Isolate or remove GST-specific antibodies This GST resin consists of purified glutatione S-transferase protein from Schistosoma japonicum that has been immobilized on crosslinked 6% beaded agarose. Antibody molecules in that cross-react with the GST portion of a recombinant
  1. 1
  2. 2
  3. 3
  4. 4
  5. >