Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Refine Browse Results
Browse Results: 13 item(s)
You're Browsing:Nucleic Acid Labeling
  1. 1
1 GE* Healthcare Amersham* CyDye* Fluorescent Nucleotides
For labeling DNA and RNA probes especially for multicolor analysis in microarrays, FISH, chromosome identification, whole chromosome painting, karyotyping, and gene mapping. CyDye labeled nucleotides are available in a range of bright, intense colors with narrow emission bands—ideal for multiplexing
2 Available on GSA/VA Contract for Federal Government customers only. Mirus Bio Label IT* Nucleic Acid Labeling Kits
Efficient, direct, nonenzymatic labeling of DNA and RNA   ·  Kits label any DNA or RNA template—suitable for a wide range of applications  ·  One-step chemical method provides easy and consistent control of the labeling reactions  ·  Adjustable labeling
3 GE* Healthcare Amersham* ECL Direct* Nucleic Acid Labeling/Detection System
For fast and easy detection of medium-to-high-target amounts   ·  Consistent results combining strong signals with very low backgrounds  ·  No need to strip the blot before reprobing: saves time, reduces membrane damage, and minimizes exposure to hazardous chemicals  ·  Less
4 GE* Healthcare illustra* CyScribe* GFX* Purification Kit
Allows rapid purification of CyDye* labeled cDNA probes   ·  Provides efficient removal of unincorporated CyDye label and primers from labeling reactions with superior yields of labeled cDNA probe  ·  Excellent for the purification of cDNA labeled by either
5 GE* Healthcare Amersham* Rediprime* II DNA Labeling System
Pre-mixed, individually dispensed, random-prime reactions, freeze-dried for ambient stability Simply add substrate and [a-32P]dCTP. As little as 25ng substrate DNA can be used.   ·  Ambient temperature-stable reactions with a color check for the
6 GE* Healthcare Amersham* Megaprime* DNA Labeling System
For labeling DNA probes to specific activities >1 x 109dpm/µg in 10 minutes Megaprime* DNA Labeling Systems are used for rapid, random-prime labeling from as little as 25ng substrate DNA.   ·  Features cloned Klenow fragment and random nonamers
7 GE* Healthcare Ready-to-Go* DNA Labeling Beads (-dCTP)
Ambient-temperature-stable, single-dose reaction spheres for labeling DNA to specific activities >109dpm/µg in as little as 5 min. using labeled dCTP   ·  Beads are predispensed in individual reaction tubes to eliminate the potential for cross-contamination  ·  Beads
8 GE* Healthcare ProbeQuant* G-50 Micro Columns
For rapid purification of labeled DNA (>20 bases) from unincorporated labeled nucleotides in a volume of 25 to 50µL using spin-column chromatography   ·  Prepacked with Sephadex* G-50 DNA, and designed for use in a microcentrifuge  ·  Requires less than
9 Marligen Vantage* microRNA Labeling Kit
Utilizes a proprietary labeling technology to provide highly sensitive detection of endogenous microRNAs Labels RNA in just two steps. The 3' end of the RNA is poly(A) tailed, followed by the ligation of biotinylated 3DNA* dendrimers containing 15 biotins to provide signal amplification.
10 GE* Healthcare Amersham* CyDye* Post-Labeling Reactive Dye Packs
Reactive CyDye fluors specifically optimized for nucleic acid microarray labeling   ·  High reactivity Cy*3 and Cy*5 dyes: 40,000pmol reactive dye per vial, at >75% reactive dye content  ·  Each vial is sufficient for one labeling reaction of up
11 GE* Healthcare AlkPhos Direct* Hybridization Buffer for Direct Labeling System
Fast and convenient system for all high-sensitivity applications using DNA, RNA, or oligonucleotide probes   ·  Hybridization stringency can be controlled by elevating temperatures up to 75°C (167°F), or by varying salt concentration  ·  Detection of 60 fg target
12 GE* Healthcare Amersham* HyPer5* Microarray Dye
For the generation of microarray probes by either post labeling (HyPer5 NHS reactive dye) or direct enzymic incorporation with HyPer5 dCTP in gene expression or aCGH experiments HyPer5 dye is a new fluorescent dye for microarray applications that combines exceptional photo-and-ozone stability with highly
13 GE* Healthcare Sonicated DNAs for Hybridization Studies
Lyophilized DNAs for reducing hybridization background caused by binding of labeled probe to non-specific sites on membranes Sonicated Salmon Sperm DNA Sonicated Calf Thymus DNA
  1. 1