Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Print Email
75-004-510, -509
View Image Gallery (3) Image(s) are representative of the product Group
  • 75-004-510, -509
Available on GSA/VA Contract for Federal Government customers only. Thermo Scientific™ Sorvall™ ST 40 Centrifuge Series
General-purpose 3L centrifuges for everyday sample preparations
Required Accessories: Rotor (sold separately)
Easy To Use
  • Intuitive controls simplify operation and provide detailed information on a wide range of operating parameters and processing status for optimal performance
  • Motorized lid latch enables one finger downward motion for easiest closing and locking of centrifuge lid, even on tall benches
Auto-Lock™ III Rotor System
  • Secure locking system allows easy push-button installation and exchange of rotors
  • Saves time switching between applications
  • Easy access to rotor chamber for quick cleaning, a healthier working environment and longer unit life
ClickSeal™ Bucket Sealing System
  • Biocontainment solution seals with a snap; eliminates screw caps and complicated clips
  • Glove-friendly, one-handed open/close capability
  • Certified by CAMR* in Porton Down, UK
SMARTSpin™ Technology
  • Advanced rotor management system maximizes acceleration, braking and residual load imbalance for each rotor and bucket option
  • Optimizes safety and improves separations
View Specifications
Images for Available on GSA/VA Contract for Federal Government customers only. Thermo Scientific™ Sorvall™ ST 40 Centrifuge Series
  • 75-004-510, -509
75-004-510, -509
75-004-510, -509
(4 item(s) in product Group)
  • Items
  • Specifications
Catalog No. Description Price per Unit Quantity & Availability Add Item(s)
75-004-510 Available on GSA/VA Contract for Federal Government customers only. 
THERMO SCIENTIFIC
No.:75004510
ST 40, Ventilated, 120V 60Hz
Each for $7,274.00
Check Availability
Add
to Cart
(0) in My Cart
75-004-509 Available on GSA/VA Contract for Federal Government customers only. 
THERMO SCIENTIFIC
No.:75004509
ST 40, Ventilated, 230V 50/60Hz
Each for $7,274.00
Check Availability
Add
to Cart
(0) in My Cart
75-004-525 Available on GSA/VA Contract for Federal Government customers only. 
THERMO SCIENTIFIC
No.:75004525
ST 40R, Refrigerated, 120V 60Hz
Each for $10,274.00
Check Availability
Add
to Cart
(0) in My Cart
75-004-524 Available on GSA/VA Contract for Federal Government customers only. 
THERMO SCIENTIFIC
No.:75004524
ST 40R, Refrigerated, 230V 50/60Hz
Each for $10,274.00
Check Availability
Add
to Cart
(0) in My Cart
Add Items to Cart
Description
Easy To Use
  • Intuitive controls simplify operation and provide detailed information on a wide range of operating parameters and processing status for optimal performance
  • Motorized lid latch enables one finger downward motion for easiest closing and locking of centrifuge lid, even on tall benches
Auto-Lock™ III Rotor System
  • Secure locking system allows easy push-button installation and exchange of rotors
  • Saves time switching between applications
  • Easy access to rotor chamber for quick cleaning, a healthier working environment and longer unit life
ClickSeal™ Bucket Sealing System
  • Biocontainment solution seals with a snap; eliminates screw caps and complicated clips
  • Glove-friendly, one-handed open/close capability
  • Certified by CAMR* in Porton Down, UK
SMARTSpin™ Technology
  • Advanced rotor management system maximizes acceleration, braking and residual load imbalance for each rotor and bucket option
  • Optimizes safety and improves separations
Space-Saving Design
  • Compact dimensions optimize work area and ergonomic height and curved design ensure easy sample loading and unloading and easy cleaning
Outstanding Energy Efficiency
  • Up to 40% energy consumption savings on industry standard protocols, such as standard blood separation or conical tubes processing
Applications: Cell Culture, Blood Processing
Ventilated Model ST 40
  • Dimensions: 26.4L x 22.2W x 14.2 in.H [35.4 in.H open] (67 x 56.5 x 36cm [90cm open])
  • Net weight: 189.6 lb. (86kg)
Refrigerated Model ST 40R
  • CFC-free refrigeration system
  • Temperature Range: -10° to +40°C
  • Pre-Cooling Function with direct button
  • 26.4L x 29.3W x 14.2 in.H [35.4 in.H open] (67 x 74.5 x 36cm [90cm open])
  • Net weight: 255.7 lb. (116kg)
Specifications & Ordering Information:
A wide range of rotors are available for your daily routine processing requirements. Maximize performance with lighter-weight Thermo Scientific Fiberlite™ carbon fiber rotors that offer versatility, speed and robust, corrosion-free design. All rotors and adapters are sold separately.
Certifications: CSA, Certified Biosafety, CE marked, UL listed
Compliance: IVD Compliant, IEC 61010-1, IEC 61010-2-202, IEC 61010-2-101
Specifications
Capacity 3L (4 x 750mL)
DisplayDigital
ControlMicroprocessor
Max. Speed15,200rpm
Max. RCF25,000 to 25,999 x G
Timer 9 hr., 99 min. plus HOLD
Memory6 programs, direct access
Other FeaturesAuto-Lock III rotor locking system, SMARTspin imbalance detection system, Certified ClickSeal biocontainment system, language selection, 9 acceleration/10 braking profiles, motorized lid latch
MotorDirect, brushless induction low-profile motor
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your 9-digit account number, or become a registered user on fishersci.com by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service at 1-800-766-7000.