Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Print Email
Drucker 853VES Series Centrifuges
Drucker™ Horizon™ Performance Series Model 853VES Plasmafuge-12
Medium capacity 12-place horizontal centrifuge with digital controls
Test tubes: Holds 5-7mL test tubes up to a 17 x 75mm and 10mL test tubes up to 17 x 100mm
Electronic timer: With display, selectable from 1 to 99 min; with HOLD feature (±1%)
Electronic control: With display, allows running speed to be easily set by rpm or rcp
Force: Up to 3800xG (depending on Rotor Selection)
Speed:
  • Standard: up to 4000rpm
  • Performance Plus: up to 5000rpm
Minimum acceleration/decelerationtime: 15 sec.
Memory: Up to 10 memory settings
View Specifications
Images for Drucker™ Horizon™ Performance Series Model 853VES Plasmafuge-12
  • Drucker 853VES Series Centrifuges
Drucker 853VES Series Centrifuges
Drucker 853VES Series Centrifuges
(2 item(s) in product Group)
  • Items
  • Specifications
Catalog No. Description Price per Unit Quantity & Availability Add Item(s)
22-029-422 853VES Performance Plus
Accessories
Each for $3,344.77
Check Availability
Add
to Cart
(0) in My Cart
22-029-421 853VES Standard
Accessories
Each for $2,738.20
Check Availability
Add
to Cart
(0) in My Cart
Description
Test tubes: Holds 5-7mL test tubes up to a 17 x 75mm and 10mL test tubes up to 17 x 100mm
Electronic timer: With display, selectable from 1 to 99 min; with HOLD feature (±1%)
Electronic control: With display, allows running speed to be easily set by rpm or rcp
Force: Up to 3800xG (depending on Rotor Selection)
Speed:
  • Standard: up to 4000rpm
  • Performance Plus: up to 5000rpm
Minimum acceleration/decelerationtime: 15 sec.
Memory: Up to 10 memory settings
Specifications & Ordering Information:
H x W x D: 8.5 x 12.5 x 15.5 in. (23 x 32 x 39cm). Weight: 30 lb. (13.7kg). 115/230V (±10%) 50/60Hz, 4A (resettable), 200w max.
Compliance: ETL Compliant to ANSI™/UL Standards; CE Marked