Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Print Email
22-024-981
View Image Gallery (4) Image(s) are representative of the product Group
  • 22-024-981
  • 22-024-982
  • 22-024-982
  • 22-024-982
Smith Medical Saf-T-Lance™ Safety Lancets
Retractable safety technology for capillary blood sampling
Automatic retraction reduces needlestick injuries and cross-contamination exposure while providing a quick, gentle puncture. Needle is shielded before and after use.
  • Built-in needle depth control provides for consistent and accurate sampling results
  • Six different flow models provide flexibility for sampling applications
  • Proprietary spring action provides quick, gentle puncture
View Specifications
Images for Smith Medical Saf-T-Lance™ Safety Lancets
  • 22-024-981
  • 22-024-982
  • 22-024-982
  • 22-024-982
22-024-981
22-024-981
(5 item(s) in product Group)
  • Items
  • Specifications
Catalog No. Description Price per Unit Quantity & Availability Add Item(s)
22-024-971
Smith Medical
No.:1020
Blue; Low Flow; 25 Gauge
Case of 3600 for $1,356.14
Check Availability
Add
to Cart
(0) in My Cart
22-024-981
SMITH MEDICAL
No.:1008
Yellow; High Flow; 18 Gauge
Case of 3600 for $1,356.14
Check Availability
Add
to Cart
(0) in My Cart
22-025-003
SMITH MEDICAL
No.:1005
Purple; Maximum Flow Blade
Case of 3600 for $1,128.90
Check Availability
Add
to Cart
(0) in My Cart
22-024-982
SMITH MEDICAL
No.:1014
Green; Normal Flow; 21 Gauge
Case of 3600 for $1,356.14
Check Availability
Add
to Cart
(0) in My Cart
22-025-005
SMITH MEDICAL
No.:1028
Pink; Pediatric Blade
N/A N/A N/A
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
Add Items to Cart
Description
  • Built-in needle depth control provides for consistent and accurate sampling results
  • Six different flow models provide flexibility for sampling applications
  • Proprietary spring action provides quick, gentle puncture