Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Print Email
View Image Gallery (2) Image(s) are representative of the product Group
  • acros
Sodium tungstate dihydrate, 99+%, ACS reagent, ACROS Organics™
Sodium tungstate dihydrate
ACS reagent
99+%
Application: Oxidation reagent
Na2O42H2O
Na2WO4·2H2O
F.W.: 329.85
[10213-10-2]
View Specifications
Images for Sodium tungstate dihydrate, 99+%, ACS reagent, ACROS Organics™
  • acros
(3 item(s) in product Group)
  • Items
  • Specifications
  • Physical Properties
  • Safety and Handling
Catalog No. Description Price per Unit Quantity & Availability Add Item(s)
AC424471000
ACROS Organics
No.:424471000
100g; Glass bottle
MSDS
EA for $82.70
Check Availability
Add
to Cart
(0) in My Cart
AC424470050
ACROS Organics
No.:424470050
5g; Glass bottle
MSDS
Each for $30.20
Check Availability
Add
to Cart
(0) in My Cart
AC424475000
ACROS Organics
No.:424475000
500g; Glass bottle
MSDS
Each for $255.70
Check Availability
Add
to Cart
(0) in My Cart
Add Items to Cart
Product Specifications
Insoluble Matter0.01% max.
Sulfate100ppm max.
Molybdenum [Mo]10ppm max.
Physical FormWhite to slightly yellow crystalline powder or flakes
Chlorine [Cl]50ppm max.
Heavy Metals and Iron [as Pb]0.001% max.
Titratable Free Base0.02 meq/g max.
Gravimetry99.0 to 101.0%
Solubility in water: 730g/L (20°C). Other solubilities: insoluble in alcohol and acids
Warning
GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.