Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Print Email
A-78100001
Available on GSA/VA Contract for Federal Government customers only. Thermo Scientific™ Finesse™ 325 Manual Microtome
Manual rotary microtome for routine paraffin sectioning; lightly balanced flywheel is designed to prevent repetitive stress injury.
Includes: Fine orienting head (Mfr. No. 0325-105); Standard quick release specimen clamp (Mfr. No. 0303); Audible and visual specimen travel alarm/indicator; Organizer Tray (Mfr. No., A7813000); Debris tray (Mfr. No. A78130003); Dust cover (Mfr. No. AP14325); Microtome oil (Mfr. No. 1520/1); Operator manual (specify language) Mfr. No. A78110100; Service Record Book (Mfr. No. A78410106)
Note: Contents, Packaging, and Quantities of items listed are subject to change without prior notice.
  • Supplied with a quick-release cassette clamp and x-y orientation head as standard
  • Disposable blades or standard microtome knives can be used with this unit (with the appropriate knife holder)
  • Modern Accu-Feed System provides high-quality, reproducible sections throughout the length of specimen travel
  • Accu-Feed specimen advance is selectable in 1µm steps from 1 to 30µm
  • Specimen tilt / orientation is ±5 degrees using X-Y micro adjustment
  • Precision response, free-running flywheel with ergonomic design
  • Removable deep debris tray for easy cleaning
View Specifications
Images for Available on GSA/VA Contract for Federal Government customers only. Thermo Scientific™ Finesse™ 325 Manual Microtome
  • A-78100001
A-78100001
A-78100001
(4 item(s) in product Group)
  • Items
  • Specifications
Catalog No. Description Price per Unit Quantity & Availability Add Item(s)
A78100001 Available on GSA/VA Contract for Federal Government customers only. 
Thermo Scientific
No.:A78100001
Microtome only
Each for $7,400.00
Check Availability
Add
to Cart
(0) in My Cart
A78100001L Available on GSA/VA Contract for Federal Government customers only. 
Thermo Scientific
No.:A78100001L
Low Profile Blade Holder Package
Each for $9,500.00
Check Availability
Add
to Cart
(0) in My Cart
A78100001H Available on GSA/VA Contract for Federal Government customers only. 
Thermo Scientific
No.:A78100001H
High profile blade holder package
Each for $9,400.00
Check Availability
Add
to Cart
(0) in My Cart
A78100001K Available on GSA/VA Contract for Federal Government customers only. 
Thermo Scientific
No.:A78100001K
Universal Blade Holder Package
Each for $9,400.00
Check Availability
Add
to Cart
(0) in My Cart
Add Items to Cart
Description
  • Supplied with a quick-release cassette clamp and x-y orientation head as standard
  • Disposable blades or standard microtome knives can be used with this unit (with the appropriate knife holder)
  • Modern Accu-Feed System provides high-quality, reproducible sections throughout the length of specimen travel
  • Accu-Feed specimen advance is selectable in 1µm steps from 1 to 30µm
  • Specimen tilt / orientation is ±5 degrees using X-Y micro adjustment
  • Precision response, free-running flywheel with ergonomic design
  • Removable deep debris tray for easy cleaning
Specifications & Ordering Information:
Specify Knife Holder at the time of order. Blade holders/Knife holders are not included with microtome unless ordering package.
Compliance: CE marked (IVDD compliant)
Specifications
DimensionsD x W x H: 19.7 x 16.1 x 11.4 in. (50 x 41 x 29cm)
Weight88 lb. (40kg)
Section Thickness Range 0.5 to 60µm
Section Thickness Range Increments0.5 to 2µm in 0.5µm increments
2 to 10µm in 1µm increments
10 to 20µm in 2µm increments
20 to 60µm in 5µm increments
Horizontal Overall Specimen Feed 28mm
Vertical Specimen Stroke 64mm
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your 9-digit account number, or become a registered user on fishersci.com by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service at 1-800-766-7000.