Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Print Email
Available on GSA/VA Contract for Federal Government customers only. Thermo Scientific™ PHARM-TROL Count Control Particle Standards
For USP <788> and <789> verification
PHARM-TROL contains NIST traceable size standards with a measured particle count. The product was developed for manufactures of parenteral drugs and ophthalmic solutions seeking interim verification of USP <788> and <789> verification (Particulate Matter in Injections and Particulate Matter in Ophthalmic Solutions). The particle diameters are traceable to the Standard Meter through the National Institute of Standards and Technology (NIST).
PHARM-TROL is prepared with the same exacting procedures as the USP Count Reference Standard and provides process control data for particle counting consistency and verification of size calibration.
Applications:Particle count control for USP <788> and <789> verification
View Specifications
Images for Available on GSA/VA Contract for Federal Government customers only. Thermo Scientific™ PHARM-TROL Count Control Particle Standards
(2 item(s) in product Group)
  • Items
  • Specifications
Catalog No. Description Price per Unit Quantity & Availability Add Item(s)
09-980-235 Available on GSA/VA Contract for Federal Government customers only. 
Thermo Scientific
No.:CS-PK
15µm; 6 x 25mL bottles
N/A N/A N/A
Price File Not Open 
09-980-236 Available on GSA/VA Contract for Federal Government customers only. 
Thermo Scientific
No.:CS-BX
15µm; 20 x 25mL bottles
N/A N/A N/A
Price File Not Open 
Applications: Particle count control for USP <788> and <789> verification
Compliance: NIST traceable
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your 9-digit account number, or become a registered user on fishersci.com by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service at 1-800-766-7000.