Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Print Email
75-004-241
View Image Gallery (3) Image(s) are representative of the product Group
  • 75-004-241
  • 75004381
Available on GSA/VA Contract for Federal Government customers only. Thermo Scientific™ Sorvall™ ST 16 Centrifuge Series
Ideal for routine processing
The Thermo Scientific Sorvall ST 16 general purpose centrifuge series is perfect for everyday sample processing, including clinical protocols, cell culture applications and microplate processing. The refrigerated version is ideal for handling temperature-sensitive sample processing between -10° and +40°C.
Required Accessories: Rotor (sold separately)
Easy To Use
  • Intuitive controls simplify operation and provide detailed information on a wide range of operating parameters and processing status for optimal performance
  • Motorized lid latch enables one finger downward motion for easiest closing and locking of centrifuge lid, even on tall benches
Auto-Lock™ III Rotor System
  • Secure locking system allows easy push-button installation and exchange of rotors
  • Saves time switching between applications
  • Easy access to rotor chamber for quick cleaning, a healthier working environment and longer unit life
ClickSeal™ Bucket Sealing System
  • Biocontainment solution seals with a snap; eliminates screw caps and complicated clips
  • Glove-friendly, one-handed open/close capability
  • Certified by CAMR™ in Porton Down, UK
SMARTSpin™ Technology
  • Advanced rotor management system maximizes acceleration, braking and residual load imbalance for each rotor and bucket option
  • Optimizes safety and improves separations
View Specifications
Images for Available on GSA/VA Contract for Federal Government customers only. Thermo Scientific™ Sorvall™ ST 16 Centrifuge Series
  • 75-004-241
  • 75004381
75-004-241
75-004-241
(4 item(s) in product Group)
  • Items
  • Specifications
Catalog No. Description Price per Unit Quantity & Availability Add Item(s)
75-004-241 Available on GSA/VA Contract for Federal Government customers only. 
THERMO SCIENTIFIC
No.:75004241
ST 16; Ventilated; 120V 60Hz
Each for $4,550.00
Check Availability
Add
to Cart
(0) in My Cart
75-004-240 Available on GSA/VA Contract for Federal Government customers only. 
THERMO SCIENTIFIC
No.:75004240
ST 16; Ventilated; 230V 50/60Hz
Each for $4,550.00
Check Availability
Add
to Cart
(0) in My Cart
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
75-004-381 Available on GSA/VA Contract for Federal Government customers only. 
THERMO SCIENTIFIC
No.:75004381
ST 16R; Refrigerated; 120V 60Hz
Each for $6,710.00
Check Availability
Add
to Cart
(0) in My Cart
75-004-380 Available on GSA/VA Contract for Federal Government customers only. 
THERMO SCIENTIFIC
No.:75004380
ST 16R; Refrigerated; 230V 50/60Hz
Each for $6,710.00
Check Availability
Add
to Cart
(0) in My Cart
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
Add Items to Cart
Description
Easy To Use
  • Intuitive controls simplify operation and provide detailed information on a wide range of operating parameters and processing status for optimal performance
  • Motorized lid latch enables one finger downward motion for easiest closing and locking of centrifuge lid, even on tall benches
Auto-Lock™ III Rotor System
  • Secure locking system allows easy push-button installation and exchange of rotors
  • Saves time switching between applications
  • Easy access to rotor chamber for quick cleaning, a healthier working environment and longer unit life
ClickSeal™ Bucket Sealing System
  • Biocontainment solution seals with a snap; eliminates screw caps and complicated clips
  • Glove-friendly, one-handed open/close capability
  • Certified by CAMR™ in Porton Down, UK
SMARTSpin™ Technology
  • Advanced rotor management system maximizes acceleration, braking and residual load imbalance for each rotor and bucket option
  • Optimizes safety and improves separations
Space-Saving Design
  • Compact dimensions optimize work area and ergonomic height and curved design ensure easy sample loading and unloading and easy cleaning
  • Outstanding Energy Efficiency
    • Up to 40% energy consumption savings on industry standard protocols, such as standard blood separation or conical tubes processing
    Applications: Cell Culture, Blood Processing
    Ventilated Model ST 16
    • Dimensions: 23.8L x 17.3W x 14.2 in.H [34.3 in.H open] (60.5 x 44 x 36cm [87cm open])
    • Net weight: 126.8 lb. (57.5kg)
    Refrigerated Model ST 16R
    • CFC-free refrigeration system
    • Temperature Range: -10° to +40°C
    • Pre-Cooling Function with direct button
    • 23.8L x 24.5W x 14.2 in.H [34.3 in.H open] (60.5 x 62.3 x 36cm [87cm open])
    • Net weight: 201.7 lb. (91.5kg)
    Specifications & Ordering Information:
    A wide range of rotors are available for your daily routine processing requirements. Maximize performance with lighter-weight Thermo Scientific Fiberlite™ carbon fiber rotors that offer versatility, speed and robust, corrosion-free design. All rotors and adapters are sold separately.
    Certifications: UL listed, CSA certified, CE marked, Certified Biosafety
    Compliance: IVD Compliant, IEC 61010-1, IEC 61010-2-202, IEC 61010-2-101
    Specifications
    Capacity1.6L (4 x 400mL)
    ControlMicroprocessor
    DisplayDigital
    Max. Speed 15,200rpm
    Max. RCF25,000 to 25,999 x G
    Memory6 programs, direct access
    Timer9 hr., 99 min. plus HOLD
    Other FeaturesAuto-Lock III rotor locking system, SMARTspin imbalance detection system, Certified ClickSeal biocontainment system, language selection, 9 acceleration/10 braking profiles, motorized lid latch
    MotorDirect, brushless induction low-profile motor
    Noise<61dBA
    This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your 9-digit account number, or become a registered user on fishersci.com by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service at 1-800-766-7000.