Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Print Email
domnick hunter HIGH FLOW TETPOR II Air/Gas Filters
View Image Gallery (2) Image(s) are representative of the product Group
  • domnick hunter HIGH FLOW TETPOR II Air/Gas Filters
  • domnick hunter HIGH FLOW TETPOR II Air/Gas Filters
Available on GSA/VA Contract for Federal Government customers only. domnick hunter HIGH FLOW TETPOR II Air/Gas Filters
Provide full retention of bacteria, viruses, and particulate in process air and gas streams, with optimum efficiency, flow rate, and strength, and zero fiber shedding
  • Optimum pleat configuration
  • Integrity testable by all methods
  • Absolute rated filtration
  • High flow rates and low pressure drops
  • Steam sterilizable up to 225 cycles at 142°C
  • Extremely robust, state-of-the-art PTFE membrane technology
  • Available in 10 in. formats and Demi cartridges
  • All cartridges integrity tested by pressure decay and aerosol challenge methods
View Specifications
Images for Available on GSA/VA Contract for Federal Government customers only. domnick hunter HIGH FLOW TETPOR II Air/Gas Filters
  • domnick hunter HIGH FLOW TETPOR II Air/Gas Filters
  • domnick hunter HIGH FLOW TETPOR II Air/Gas Filters
domnick hunter HIGH FLOW TETPOR II Air/Gas Filters
(8 item(s) in product Group)
  • Items
  • Specifications
Description Catalog No. Availability Quantity Price
5in.
14-950-423 Available on GSA/VA Contract for Federal Government customers only. 
Domnick Hunter
No.:ZHFT/KC
N/AYou must be logged in and have a valid account number to view item availability   Each for $273.76
(0) in My Cart
10in.
14-950-420 Available on GSA/VA Contract for Federal Government customers only. 
Domnick Hunter
No.:ZHFT/1C
N/AYou must be logged in and have a valid account number to view item availability   Each for $499.00
(0) in My Cart
20in.
14-950-421 Available on GSA/VA Contract for Federal Government customers only. 
Domnick Hunter
No.:ZHFT/2C
N/AYou must be logged in and have a valid account number to view item availability   Each for $943.29
(0) in My Cart
1.5in.
14-950-422 Available on GSA/VA Contract for Federal Government customers only. 
Domnick Hunter
No.:ZHFT/3C
N/AYou must be logged in and have a valid account number to view item availability   Each for $1,422.00
(0) in My Cart
1.5in.
14-950-427 Available on GSA/VA Contract for Federal Government customers only. 
Domnick Hunter
No.:ZHFT/DW
N/AYou must be logged in and have a valid account number to view item availability   Pack of 3 for $503.00
(0) in My Cart
2.5in.
14-950-426 Available on GSA/VA Contract for Federal Government customers only. 
Domnick Hunter
No.:ZHFT/CW
N/AYou must be logged in and have a valid account number to view item availability   Pack of 3 for $557.50
(0) in My Cart
2.5in.
14-950-425 Available on GSA/VA Contract for Federal Government customers only. 
Domnick Hunter
No.:ZHFT/BZ
N/AYou must be logged in and have a valid account number to view item availability   Pack of 3 for $538.00
(0) in My Cart
5in.
14-950-424 Available on GSA/VA Contract for Federal Government customers only. 
Domnick Hunter
No.:ZHFT/AZ
N/AYou must be logged in and have a valid account number to view item availability   Pack of 3 for $660.50
(0) in My Cart
Add Items to Cart
Description
  • Optimum pleat configuration
  • Integrity testable by all methods
  • Absolute rated filtration
  • High flow rates and low pressure drops
  • Steam sterilizable up to 225 cycles at 142°C
  • Extremely robust, state-of-the-art PTFE membrane technology
  • Available in 10 in. formats and Demi cartridges
  • All cartridges integrity tested by pressure decay and aerosol challenge methods
Applications:
  • Suitable for all sterile gas applications, including fermentation inlet and off gas streams, venting, lyophilizers, autoclave vacuum breaks, and blow-fill-seal equipment
  • Also suitable for particle-free air applications in the electronics industry
Compliance:
  • Biological Safety: Conforms to current USP Plastics Class VI - 121°C
  • Fully validated to ASTM F838-83
  • Fully validated to aerosol bacterial and viral challenge
  • Fully validated as sterilizing grade filter cartridges, for compressed air and gas applications
  • Exceed liquid bacterial challenge levels as recommended by ASTM
  • Also validated by aerosol bacterial and MS-2 Coliphage challenge testing
  • Full validation guide is available upon request
Specifications
MembraneExpanded PTFE
Upstream SupportsExpanded polypropylene and nonwoven polypropylene
Downstream SupportExpanded polypropylene
Core316L stainless steel (HIGH FLOW TETPOR II) or Polypropylene (Demi HIGH FLOW TETPOR II)
SleeveHeat-stabilized polypropylene
EndcapsHeat-stabilized polypropylene (with integral polysulfone support ring on 226 style)
Maximum Differential PressureIn direction of flow (outside to in) 3.5 barg (50 psig) at 142°C (288°F)
Maximum Recommended Continuous Inlet Air Temperature60°C (140°F) Note: HIGH FLOW TETPOR II cartridges can be used as WFI vents in heated housings if changed on a 4 to 6 monthly basis.
Maximum Steam Temperature142°C (288°F)
SterilizationCan be in-situ steam sterilized for up to 225 cycles at 142°C (288°F)
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your 9-digit account number, or become a registered user on fishersci.com by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service at 1-800-766-7000.