Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Print Email
View Image Gallery (6) Image(s) are representative of the product Group
  • Int several tabs
Green Product Thermo Scientific* 1300 Series A2 Class II, Type A2 Biological Safety Cabinet Packages
Designed for working with biological agents at Biosafety Levels 1–3
Thermo Scientific 1300 Series A2 features advancements in brushless DC motor technology for dramatically improved energy efficiency, safety performance, and reliability. SmartClean offers a window cleaning position that provides the user access to interior of window sash. SmartPort provides a clean and easy way to organize the interior of the 1300 Series A2, with confidence due to our negative pressure dual-side wall.
  • Energy efficient design helps labs save money over the entire life of the cabinet
  • Reduce operating costs up to 75% over traditional biological safety cabinets with AC motors
  • Increase comfort and encourage safe working habits with the sloped front, bright workspace, and low noise level
  • Maximize safety through our innovative SmartFlow* design with Digital Airflow Verification
View Specifications
Images for Green Product Thermo Scientific* 1300 Series A2 Class II, Type A2 Biological Safety Cabinet Packages
  • Int several tabs
(17 item(s) in product Group)
  • Items
  • Specifications
Description Catalog No. Availability Quantity Price
Width: 72 in.; Stainless-steel package; 120V 50/60Hz
13-261-313
Thermo Scientific
No.:1337
N/AYou must be logged in and have a valid account number to view item availability   Each for $11,730.00
(0) in My Cart
Width: 48 in.; 328 to 361cfm
13-261-222
Thermo Scientific
No.:1375
N/AYou must be logged in and have a valid account number to view item availability   Each for $9,740.00
(0) in My Cart
Width: 60 in.; 10 in. window; Coated interior cabinet walls; 120V 50/60Hz
13-261-336
Thermo Scientific
No.:1369
N/AYou must be logged in and have a valid account number to view item availability   Each for $10,640.00
(0) in My Cart
Width: 72 in.; Coated interior cabinet walls; 120V 50/60Hz
13-261-315
Thermo Scientific
No.:1397
N/AYou must be logged in and have a valid account number to view item availability   Each for $11,280.00
(0) in My Cart
Window: 48 x 10 in.
13-261-304
THERMO SCI ENCLOSURES
No.:1385
N/AYou must be logged in and have a valid account number to view item availability   Each for $9,310.00
(0) in My Cart
Width: 72 in.; 492 to 541cfm
13-261-223
Thermo Scientific
No.:1377
N/AYou must be logged in and have a valid account number to view item availability   Each for $11,730.00
(0) in My Cart
Width: 36 in.; 8 in. window; Stainless-steel package; 120V 50/60Hz
13-261-342
Thermo Scientific
No.:1342
N/AYou must be logged in and have a valid account number to view item availability   Each for $9,200.00
(0) in My Cart
Width: 60 in.; 8 in. window; Coated interior cabinet walls; 120V 50/60Hz
13-261-344
Thermo Scientific
No.:1351
N/AYou must be logged in and have a valid account number to view item availability   Each for $10,640.00
(0) in My Cart
Width: 60 in.; 10 in. window; Stainless-steel package; 120V 50/60Hz
13-261-338
Thermo Scientific
No.:1371
N/AYou must be logged in and have a valid account number to view item availability   Each for $11,090.00
(0) in My Cart
Width: 36 in.; 8 in. window; Coated interior cabinet walls; 120V 50/60Hz
13-261-340
Thermo Scientific
No.:1340
N/AYou must be logged in and have a valid account number to view item availability   Each for $8,770.00
(0) in My Cart
Window: 72 x 10 in.
13-261-306
THERMO SCI ENCLOSURES
No.:1387
N/AYou must be logged in and have a valid account number to view item availability   Each for $11,280.00
(0) in My Cart
Width: 36 in.; 10 in. window; Coated interior cabinet walls; 120V 50/60Hz
13-261-332
Thermo Scientific
No.:1321
N/AYou must be logged in and have a valid account number to view item availability   Each for $8,770.00
(0) in My Cart
Width: 48 in.; Coated interior cabinet walls; 120V 50/60Hz
13-261-314
Thermo Scientific
No.:1395
N/AYou must be logged in and have a valid account number to view item availability   Each for $9,310.00
(0) in My Cart
Width: 36 in.; 10 in. window; Stainless-steel package; 120V 50/60Hz
13-261-334
Thermo Scientific
No.:1323
N/AYou must be logged in and have a valid account number to view item availability   Each for $9,200.00
(0) in My Cart
Width: 48 in.; Stainless-steel package; 120V 50/60Hz
13-261-312
Thermo Scientific
No.:1335
N/AYou must be logged in and have a valid account number to view item availability   Each for $9,740.00
(0) in My Cart
Width: 60 in.; 8 in. window; Stainless-steel package; 120V 50/60Hz
13-261-346
Thermo Scientific
No.:1353
N/AYou must be logged in and have a valid account number to view item availability   Each for $11,090.00
(0) in My Cart
Window W: 72 x 10 in.; Coated interior walls; 230V 50/60Hz
13261307
THERMO SCI ENCLOSURES
No.:1388
N/A N/A N/A
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
Add Items to Cart
Description
  • Energy efficient design helps labs save money over the entire life of the cabinet
  • Reduce operating costs up to 75% over traditional biological safety cabinets with AC motors
  • Increase comfort and encourage safe working habits with the sloped front, bright workspace, and low noise level
  • Maximize safety through our innovative SmartFlow* design with Digital Airflow Verification
Safety Features
  • Meet NSF/ANSI* Standard 49
  • Real-time airflow adjustments ensure inflow and downflow velocities remain steady
  • Pressure sensor filter monitoring measures total airflow across the working area
  • Proprietary front window design simplifies cleaning for a safer working environment
  • SmartFlow* Indicator adjusts airflows as filter resistance changes
  • SmartFlow visually demonstrates cabinet's ability to compensate to maintain personal protection
More Choices
  • Available in 3, 4, 5 or 6 ft. widths (91.4, 122, 152.4 or 183cm)
  • Available with either 8 or 10 in. work aperture (20.3 or 25.4cm)
  • Available in SmartCoat or stainless-steel interior
  • Easy to order package configurations include popular accessories (manual adjustable height stand, UV light, and armrest)
  • Electrical or manual adjustable height stands and Ergolign chair are also available
Certifications: NSF/ANSI 49, UL, CE
Warranty: Three years parts and labor on equipment
Specifications
TypeClass II, Type A2
CertificationNSF/ANSI 49, UL, CE
FilterH14 HEPA EN 1822, 99.995% MPPS
Max. Opening Height21 in. (53.5cm)
Load Bearing Capacity110 lb. (50kg)
Service Fixtures6 (3 on each side)