Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Print Email
HyClone™ Classical Liquid Media: RPMI 1640
System tested with Thermo Scientific HyClone sera to ensure product efficacy and homogeneity
All products are hydrated using WFI quality water and undergo 0.1µm sterile filtration
Manufactured using ISO 9001:2000-certified processes
Offered in a variety of packaging configurations, including convenient ™ six-packs
View Specifications
Alert: Hyclone Cell Culture products now a part of GE Healthcare.
Images for HyClone™ Classical Liquid Media: RPMI 1640
(12 item(s) in product Group)
  • Items
  • Specifications
Catalog No. Description Price per Unit Quantity & Availability Add Item(s)
SH3002701
GE Healthcare
No.:SH30027.01
Size: 500mL; With L-Glutamine
Each for $20.04
Check Availability
Add
to Cart
(0) in My Cart
SH30027FS
GE Healthcare
No.:SH30027FS
With L-Glutamine; 6 x 500mL
Pack of 6 for $114.23
Check Availability
Add
to Cart
(0) in My Cart
SH3002702
GE Healthcare
No.:SH30027.02
With L-Glutamine; 1000mL
Each for $32.05
Check Availability
Add
to Cart
(0) in My Cart
SH30027LS
GE Healthcare
No.:SH30027LS
With L-Glutamine; 6 x 1000mL
Pack of 6 for $182.70
Check Availability
Add
to Cart
(0) in My Cart
SH3009601
GE Healthcare
No.:SH30096.01
Without Glutamine; With Phenol Red; 500mL
MSDS
Each for $20.40
Check Availability
Add
to Cart
(0) in My Cart
SH30096FS
GE Healthcare
No.:SH30096.FS
Without L-Glutamine; 6 x 500mL
Pack of 6 for $116.28
Check Availability
Add
to Cart
(0) in My Cart
SH3009602
GE Healthcare
No.:SH30096.02
Without Glutamine; With Phenol Red; 1L
Each for $32.64
Check Availability
Add
to Cart
(0) in My Cart
SH30096LS
GE Healthcare
No.:SH30096LS
Without L-Glutamine; 6 x 1000mL
Pack of 6 for $186.05
Check Availability
Add
to Cart
(0) in My Cart
SH3025501
GE Healthcare
No.:SH30255.01
With L-Glutamine, With HEPES; 500mL
Each for $30.05
Check Availability
Add
to Cart
(0) in My Cart
SH30255FS
GE Healthcare
No.:SH30255FS
With L-Glutamine, With HEPES; 6 x 500mL
Pack of 6 for $171.28
Check Availability
Add
to Cart
(0) in My Cart
SH3025502
GE Healthcare
No.:SH30255.02
With L-Glutamine, With HEPES; 1000mL
Each for $48.08
Check Availability
Add
to Cart
(0) in My Cart
SH3060501
GE Healthcare
No.:SH30605.01
without Glutamine and without Phenol red; 500mL
Each for $29.44
Check Availability
Add
to Cart
(0) in My Cart
Add Items to Cart
Description
System tested with Thermo Scientific HyClone sera to ensure product efficacy and homogeneity
All products are hydrated using WFI quality water and undergo 0.1µm sterile filtration
Manufactured using ISO 9001:2000-certified processes
Offered in a variety of packaging configurations, including convenient ™ six-packs
Specifications & Ordering Information:
Media quantities over 10L are supplied in BioProcess Containers™ (BPC). Some products are available in convenient HyQPAK™ six-packs.