Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Print Email
anti-Cytokeratin , PE, Clone: CAM 5.2, BD
Anti-Cytokeratin (CAM 5.2) reagent has a primary reactivity with human keratin proteins that correspond to Moll's peptides No.7 and No.8, Mr 48 and 52kDa, respectively.Cytokeratin 7 and 8 are present on secretory epithelia of normal human tissue but not on stratified squamous epithelium. Anti-Cytokeratin (CAM 5.2) stains most epithelial-derived tissue, including liver, renal tubular epithelium, and hepatocellular and renal cell carcinomas. Anti-Cytokeratin (CAM 5.2) may not react with some squamous cell carcinomas.
Applications:Flow Cytometry, Intracellular Staining
View Specifications
Images for anti-Cytokeratin , PE, Clone: CAM 5.2, BD
(1 item(s) in product Group)
  • Items
  • Specifications
Catalog No. Description Price per Unit Quantity & Availability Add Item(s)
BDB347204
BD
No.:347204
50 Tests; PE
Applications: Flow Cytometry, Intracellular Staining
Recommended Storage: Refrigerate at 2-8°C