Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Print Email
06-423-2B
View Image Gallery (2) Image(s) are representative of the product Group
  • 06-423-2B
  • 1364620
Available on GSA/VA Contract for Federal Government customers only. PYREX™ Low-Actinic Media/Solution Bottles
Ideal for storing and transporting light-sensitive materials
Small Wide-Mouth Bottles
  • Graduated in mL increments
  • Permanent white enamel graduations and marking spots
  • Thread size, GL45; neck opening, 29mm ID
  • Orange autoclavable polypropylene plug-seal caps (06-414-2A) with replaceable drip-free clear pouring rings (06-414-2B)
Large Wide-Mouth Bottles
  • Orange, autoclavable linerless, one-piece polypropylene plug seal GL45 threaded cap with a drip-free pouring ring
  • Glass bead indicates full capacity line
  • Permanent white enamel graduations and marking spots
View Specifications
Alert: Bottles larger than 2L should not be used with bottle top filter units, or in other applications involving vacuum pressure, as breakage may occur. Do not tighten caps immediately after autoclaving as the vacuum resulting from cooling can cause breakage. A vented cap is not recommended for use on the 5L bottle for venting during autoclaving.
Images for Available on GSA/VA Contract for Federal Government customers only. PYREX™ Low-Actinic Media/Solution Bottles
  • 06-423-2B
  • 1364620
06-423-2B
06-423-2B
(9 item(s) in product Group)
  • Items
  • Specifications
Catalog No. Description Price per Unit Quantity & Availability Add Item(s)
06-423-2A Available on GSA/VA Contract for Federal Government customers only. 
Corning Life Sciences Glass
No.:51395100
3.38 oz. (100mL)
Accessories
Case of 4 for $113.58
Check Availability
Add
to Cart
(0) in My Cart
06-423-2BAvailable on GSA/VA Contract for Federal Government customers only. 
Corning Life Sciences Glass
No.:51395250
8.45 oz. (250mL)
Accessories
Case of 4 for $172.55
Check Availability
Add
to Cart
(0) in My Cart
06-423-2C Available on GSA/VA Contract for Federal Government customers only. 
Corning Life Sciences Glass
No.:51395500
16.9 oz. (500mL)
Accessories
Case of 4 for $203.28
Check Availability
Add
to Cart
(0) in My Cart
06-423-2D Available on GSA/VA Contract for Federal Government customers only. 
Corning Life Sciences Glass
No.:513951L
33.8 oz. (1000mL)
Accessories
Case of 4 for $233.31
Check Availability
Add
to Cart
(0) in My Cart
13-646-20 Available on GSA/VA Contract for Federal Government customers only. 
Corning Life Sciences Glass
No.:51395-25
0.84 oz. (25mL)
Accessories
Case of 4 for $95.76
Check Availability
Add
to Cart
(0) in My Cart
13-646-21 Available on GSA/VA Contract for Federal Government customers only. 
Corning Life Sciences Glass
No.:5139550
1.69 oz. (50mL)
Accessories
Case of 4 for $99.33
Check Availability
Add
to Cart
(0) in My Cart
06-423-2F Available on GSA/VA Contract for Federal Government customers only. 
Corning Life Sciences Glass
No.:51395-2L
0.52 gal. (2L)
Case of 4 for $410.03
Check Availability
Add
to Cart
(0) in My Cart
06-423-2G Available on GSA/VA Contract for Federal Government customers only. 
Corning Life Sciences Glass
No.:51395-5L
1.32 gal. (5L)
Case for $296.63
Check Availability
Add
to Cart
(0) in My Cart
06-423-2J Available on GSA/VA Contract for Federal Government customers only. 
Corning Life Sciences Glass
No.:51395-10L
2.64 gal. (10L)
Case for $351.23
Check Availability
Add
to Cart
(0) in My Cart
Add Items to Cart
Description
Small Wide-Mouth Bottles
  • Graduated in mL increments
  • Permanent white enamel graduations and marking spots
  • Thread size, GL45; neck opening, 29mm ID
  • Orange autoclavable polypropylene plug-seal caps (06-414-2A) with replaceable drip-free clear pouring rings (06-414-2B)
Large Wide-Mouth Bottles
  • Orange, autoclavable linerless, one-piece polypropylene plug seal GL45 threaded cap with a drip-free pouring ring
  • Glass bead indicates full capacity line
  • Permanent white enamel graduations and marking spots
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your 9-digit account number, or become a registered user on fishersci.com by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service at 1-800-766-7000.