Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Print Email
Thermo Scientific HyClone WFI Quality Water, Bioprocessing Scale
View Image Gallery (2) Image(s) are representative of the product Group
  • Thermo Scientific HyClone WFI Quality Water, Bioprocessing Scale
  • Thermo Scientific HyClone WFI Quality Water, Research Scale
HyClone™ Water, WFI (Water for Injection) Quality
Manufactured under ISO 9001 and cGMP guidelines
WFI quality water with the quality and reliability you have come to expect from Thermo Scientific HyClone products.
Quality:
  • Meets stringent USP or EP specifications
  • Certificate of Analysis included with product and available electronically for future reference
  • Bioprocess volumes available in industry leading animal derived component free (ADCF) film
Reliability:
  • Redundant manufacturing sites
  • Standard stock items from 500mL to 200L
Flexibility:
  • Volumes up to 1000L available as made to order
  • Customized packaging available
View Specifications
Alert: Hyclone Cell Culture products now a part of GE Healthcare.
Images for HyClone™ Water, WFI (Water for Injection) Quality
  • Thermo Scientific HyClone WFI Quality Water, Bioprocessing Scale
  • Thermo Scientific HyClone WFI Quality Water, Research Scale
Thermo Scientific HyClone WFI Quality Water, Bioprocessing Scale
Thermo Scientific HyClone WFI Quality Water, Bioprocessing Scale
(8 item(s) in product Group)
  • Items
  • Specifications
Catalog No. Description Price per Unit Quantity & Availability Add Item(s)
SH3022117
GE Healthcare
No.:SH30221.17
500mL bottle; WFI Quality Water
Each for $13.85
Check Availability
Add
to Cart
(0) in My Cart
SH3022110
GE Healthcare
No.:SH30221.10
1000mL bottle; WFI Quality Water
Each for $21.84
Check Availability
Add
to Cart
(0) in My Cart
SH30221LS
GE Healthcare
No.:SH30221LS
6 x 1000mL bottle; WFI Quality Water
Pack of 6 for $124.49
Check Availability
Add
to Cart
(0) in My Cart
SH3022124
GE Healthcare
No.:SH30221.24
10L bag; WFI Quality Water
Each for $272.80
Check Availability
Add
to Cart
(0) in My Cart
SH3022125
GE Healthcare
No.:SH30221.25
20L bag; WFI Quality Water
Each for $352.61
Check Availability
Add
to Cart
(0) in My Cart
SH3022126
GE Healthcare
No.:SH30221.26
50L bag; WFI Quality Water
Each for $1,132.06
Check Availability
Add
to Cart
(0) in My Cart
SH3022127
GE Healthcare
No.:SH30221.27
100L bag; WFI Quality Water
Each for $1,531.03
Check Availability
Add
to Cart
(0) in My Cart
SH3022128
GE Healthcare
No.:SH30221.28
200L bag; WFI Quality Water
Each for $2,291.93
Check Availability
Add
to Cart
(0) in My Cart
Add Items to Cart
Description
Quality:
  • Meets stringent USP or EP specifications
  • Certificate of Analysis included with product and available electronically for future reference
  • Bioprocess volumes available in industry leading animal derived component free (ADCF) film
Reliability:
  • Redundant manufacturing sites
  • Standard stock items from 500mL to 200L
Flexibility:
  • Volumes up to 1000L available as made to order
  • Customized packaging available
Specifications & Ordering Information:
Volumes over 1L are packaged in bioproces container bag systems. SH30221.LS is supplied in convenient HyQPAK™ six-packs.
Certifications: Manufactured under cGMP and ISO 9001 certified facilities
Recommended Storage: Store between 2° to 30°C
Specifications
US Specifications - SH part numbers; tested to USP monograph for Packaged Sterile Purified Water
Purification MethodReverse Osmosis/Distillation
AppearanceClear Liquid
SterilityMeets requirements of USP <71>
Endotoxin<0.25EU/mL
Conductivity†<5µS/cm at 25 ± 1°C
Oxidizable SubstancesNot Detected
Particulate MatterMeets requirements of USP <788>
EU Specifications - CH part numbers; tested to EP monograph for Sterilized Water for Injections
Purification MethodReverse Osmosis/Distillation
AppearanceClear Colorless Liquid
SterilityMeets requirements of EP (2.6.1) and USP <71>
Endotoxin<0.25EU/mL
Conductivity†<1.1µS/cm at 25 ± 1°C
Oxidizable SubstancesNot Detected
Particulate Contamination0.1µ final filtration
Acidity or AlkalinityPasses EP test for Sterilized Water for Injections
ChloridesPasses EP test for Sterilized Water for Injections
Nitrates<=0.2ppm
SulfatesPasses EP test for Sterilized Water for Injections
Ammonium<=0.2ppm
Calcium and MagnesiumPasses EP test for Sterilized Water for Injections
Residue on Evaporation<=0.003%
Total Organic Carbon†<500ppm
†Tested on water system day of fill
Disclaimer:
FOR FURTHER MANUFACTURING OR RESEARCH USE. NOT FOR DIAGNOSTIC OR THERAPEUTIC USE.