Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Print Email
Available on GSA/VA Contract for Federal Government customers only. Thermo Scientific™ TX-400 Rotor Adapters
For use with TX-400 Swinging Bucket Rotor
The adapters enable rotor to accept bottles up to 250mL and tubes to 100mL.
  • Maximum adapter capacity: 4 x 250mL (4 x 400mL with 400mL polypropylene bio-bottles)
  • Maximum adapter O.D. x L: 62 x 125mm (80 x 125mm with 400mL bio-bottles)
View Specifications
Images for Available on GSA/VA Contract for Federal Government customers only. Thermo Scientific™ TX-400 Rotor Adapters
(16 item(s) in product Group)
  • Items
  • Specifications
Catalog No. Description Price per Unit Quantity & Availability Add Item(s)
75-003-788 Available on GSA/VA Contract for Federal Government customers only. 
No.:75003788
175/200/225/250mL Bottle
Each for $395.00
Check Availability
Add
to Cart
(0) in My Cart
75-003-708 Available on GSA/VA Contract for Federal Government customers only. 
No.:75003708
100mL Round-bottom, Open-tube
Each for $380.00
Check Availability
Add
to Cart
(0) in My Cart
75-003-707 Available on GSA/VA Contract for Federal Government customers only. 
No.:75003707
50mL DIN Round-bottom
Each for $378.00
Check Availability
Add
to Cart
(0) in My Cart
75-003-799 Available on GSA/VA Contract for Federal Government customers only. 
No.:75003799
50mL Round-bottom
Each for $379.00
Check Availability
Add
to Cart
(0) in My Cart
75-003-683 Available on GSA/VA Contract for Federal Government customers only. 
No.:75003683
50mL Conical
Each for $390.00
Check Availability
Add
to Cart
(0) in My Cart
75-003-703 Available on GSA/VA Contract for Federal Government customers only. 
No.:75003703
30/25mL DIN Round/Flat-bottom
Each for $386.00
Check Availability
Add
to Cart
(0) in My Cart
75-003-706 Available on GSA/VA Contract for Federal Government customers only. 
No.:75003706
25mL Universal
Each for $378.00
Check Availability
Add
to Cart
(0) in My Cart
75-003-682 Available on GSA/VA Contract for Federal Government customers only. 
No.:75003682
15mL Conical
Each for $395.00
Check Availability
Add
to Cart
(0) in My Cart
75-003-794 Available on GSA/VA Contract for Federal Government customers only. 
No.:75003794
15mL Blood Collection (17 x 125mm)
Each for $378.00
Check Availability
Add
to Cart
(0) in My Cart
75-003-704 Available on GSA/VA Contract for Federal Government customers only. 
No.:75003704
15mL Round-bottom Sarstedt
Each for $388.00
Check Availability
Add
to Cart
(0) in My Cart
75-003-798 Available on GSA/VA Contract for Federal Government customers only. 
No.:75003798
14mL Conical Urine
Each for $388.00
Check Availability
Add
to Cart
(0) in My Cart
75-003-681 Available on GSA/VA Contract for Federal Government customers only. 
No.:75003681
10mL Blood Collection or 15mL Corex/Kimble
Each for $390.00
Check Availability
Add
to Cart
(0) in My Cart
75-003-680 Available on GSA/VA Contract for Federal Government customers only. 
No.:75003680
5/7mL Blood Collection
Each for $388.00
Check Availability
Add
to Cart
(0) in My Cart
75-003-793 Available on GSA/VA Contract for Federal Government customers only. 
No.:75003793
3mL RIA or Round-bottom (without cap)
Each for $382.00
Check Availability
Add
to Cart
(0) in My Cart
75-003-700 Available on GSA/VA Contract for Federal Government customers only. 
No.:75003700
1.5/2.0mL Conical Microtube
Each for $388.00
Check Availability
Add
to Cart
(0) in My Cart
75-003-825 Available on GSA/VA Contract for Federal Government customers only. 
No.:75003825
4.5/6mL Greiner Blood Tubes
Pack of 4 for $594.00
Check Availability
Add
to Cart
(0) in My Cart
Add Items to Cart
Description
  • Maximum adapter capacity: 4 x 250mL (4 x 400mL with 400mL polypropylene bio-bottles)
  • Maximum adapter O.D. x L: 62 x 125mm (80 x 125mm with 400mL bio-bottles)
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your 9-digit account number, or become a registered user on fishersci.com by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service at 1-800-766-7000.