Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Print Email
Available on GSA/VA Contract for Federal Government customers only. BD PI/RNase Staining Buffer
May be used to analyze cell cycle by flow cytometry or used with antibodies to examine the expression of proteins during the cell cycle
Propidium Iodide (PI) is a fluorescent vital dye that stains DNA and RNA. PI binds to both DNA and RNA, so the latter must be removed by digestion with ribonucleases. The content of DNA as determined by flow cytometry, can reveal useful information about the cell cycle and the proteins involved in the regulation of the cell cycle. Cells in G2 and M phases of the cell cycle have double the DNA content of those in G0 and G1 phases. Cells in S phase have a DNA content lying between these extremes.
  • PI is detected using a 562-588nm band pass filter
  • Storage Buffer: Aqueous buffered solution containing ≤0.09% sodium azide
View Specifications
Images for Available on GSA/VA Contract for Federal Government customers only. BD PI/RNase Staining Buffer
(1 item(s) in product Group)
  • Items
  • Specifications
Catalog No. Description Price per Unit Quantity & Availability Add Item(s)
BDB550825 Available on GSA/VA Contract for Federal Government customers only. 
BD
No.:550825
100mL
Each for $90.00
Check Availability
Add
to Cart
(0) in My Cart
Description
  • PI is detected using a 562-588nm band pass filter
  • Storage Buffer: Aqueous buffered solution containing ≤0.09% sodium azide
Recommended Storage: Store at 4°C. Protect from light. Do not freeze. Do not dilute or boil.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your 9-digit account number, or become a registered user on fishersci.com by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service at 1-800-766-7000.