Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Print Email
View Image Gallery (2) Image(s) are representative of the product Group
  • acros
Pivaldehyde, 97%, ACROS Organics™
Pivaldehyde
97%
Trimethylacetaldehyde
C5H10O
(CH3)3CCHO
F.W.: 86.13
[630-19-3]
View Specifications
Images for Pivaldehyde, 97%, ACROS Organics™
  • acros
(3 item(s) in product Group)
  • Items
  • Specifications
  • Physical Properties
  • Safety and Handling
Catalog No. Description Price per Unit Quantity & Availability Add Item(s)
AC368360250
ACROS Organics
No.:368360250
25g
MSDS
Each for $188.00
Check Availability
Add
to Cart
(0) in My Cart
AC368360050
ACROS Organics
No.:368360050
5g
MSDS
Each for $53.80
Check Availability
Add
to Cart
(0) in My Cart
AC368361000
ACROS Organics
No.:368361000
100g
MSDS
Each for $398.80
Check Availability
Add
to Cart
(0) in My Cart
Add Items to Cart
Product Specifications
Assay96.0% min.
Physical FormClear colorless liquid
Infrared SpectrumAuthentic
Refractive Index1.3775 to 1.3795 (20°C, 589nm)
Boiling Point: 75.0°C
Density: 0.7750g/mL
Flash Point: -15°C
Melting Point: 6.0°C
n: 1.3775 to 1.3795
Solubility in water: negligibly soluble.
Sp. gr.: 0.7750
1989
GHS Signal Word: Danger
GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Highly flammable liquid and vapor.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.