Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Print Email
03-692-180
View Image Gallery (2) Image(s) are representative of the product Group
  • 03-692-180
  • Fisherbrand Liquid Handling Products
Fisherbrand™ Bottle-Top Dispensers
Five fully adjustable models cover a dispensing range from 0.05 to 50mL
Includes: 38, 40 and 45mm adapters and an individual calibration certificate. Optional reservoirs are sold separately.
  • Precision valve mechanism ensures easy priming and minimum reagent waste, with no leakage back to the reservoir
  • Polypropylene protection sleeve surrounds the borosilicate glass barrel
  • Antidrip taps prevent accidental dispensing
  • Spring-lock provides quicker, more accurate volume selection
  • Dispenser head can be rotated 360° for convenient dispensing and volume adjustment
View Specifications
Images for Fisherbrand™ Bottle-Top Dispensers
  • 03-692-180
  • Fisherbrand Liquid Handling Products
03-692-180
03-692-180
(5 item(s) in product Group)
  • Items
  • Specifications
Catalog No. Description Price per Unit Quantity & Availability Add Item(s)
03-692-180 Range: 1 to 30mL; Increments: 1mL
Accessories
N/A N/A N/A
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
03-692-181 Range: 1 to 50mL; Increments: 1mL
Accessories
N/A N/A N/A
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
03-692-179 Range: 0.2 to 10mL; Increments: 0.2mL
Accessories
N/A N/A N/A
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
03-692-178 Range: 0.1 to 5mL; Increments: 0.1mL
Accessories
N/A N/A N/A
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
03-692-177 Range: 0.05 to 2.5mL; Increments: 0.05mL
Accessories
N/A N/A N/A
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Add Items to Cart
Description
  • Precision valve mechanism ensures easy priming and minimum reagent waste, with no leakage back to the reservoir
  • Polypropylene protection sleeve surrounds the borosilicate glass barrel
  • Antidrip taps prevent accidental dispensing
  • Spring-lock provides quicker, more accurate volume selection
  • Dispenser head can be rotated 360° for convenient dispensing and volume adjustment
  • Smooth-moving PTFE piston minimizes operator fatigue and can be removed in seconds for cleaning
  • Dispensers fit directly onto 30mm-neck bottles
  • Accuracy: ±0.3% at maximum volume
  • Repeatability: ±0.1% or better
  • Only the liquid pathway is autoclavable to 121°C (250°F)
Certifications: CE marked
Manufactured to ISO 9001:2008 quality system standards