Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Print Email
BP2956-100
View Image Gallery (2) Image(s) are representative of the product Group
  • BP2956-100
  • fisherbiotech
Puromycin Dihydrochloride, Fisher BioReagents
Puromycin Dihydrochloride
Dry, White or off-white powder
> 98 %
Inhibits protein synthesis.
C22H29N7O5·2HCl
F.W.: 544.44
[58-58-2]
View Specifications
Images for Puromycin Dihydrochloride, Fisher BioReagents
  • BP2956-100
  • fisherbiotech
BP2956-100
BP2956-100
(1 item(s) in product Group)
  • Items
  • Specifications
  • Physical Properties
  • Safety and Handling
Catalog No. Description Price per Unit Quantity & Availability Add Item(s)
BP2956-100 Clear Glass; 100mg
MSDS
Each for $282.32
Check Availability
Add
to Cart
(0) in My Cart
Product Specifications
Purity98% min.
SolubilityClear, colorless
Water< 12.0
SourceStreptomyces alboniger
Melting Point: 168° to 178°C/334.4° to 352.4°F
WARNING!
Emergency Overview
Harmful if swallowed. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Get medical attention immediately if symptoms occur. Do not induce vomiting. Do not induce vomiting. Call a physician or Poison Control Center immediately.
NFPA
Health:2
Flammability:0
Instability:0