Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Print Email
View Image Gallery (5) Image(s) are representative of the product Group
  • 14-826-87
  • 14-829-10D
  • 14-823-2E
  • 14-826-88
BD™ Tuberculin Syringes
Sterile, disposable syringe with Luer slip tip
  • Color-coded needle package and hub In Easy Peel™ package; autoclavable out of package Available with or without detachable PrecisionGlide™ needle
  • Syringe 14-823-2F accepts PrecisionGlide, Yale™, and Perfektum™ disposable and reusable needles
View Specifications
Images for BD™ Tuberculin Syringes
  • 14-826-87
  • 14-829-10D
  • 14-823-2E
  • 14-826-88
(5 item(s) in product Group)
  • Items
  • Specifications
Catalog No. Description Price per Unit Quantity & Availability Add Item(s)
14-829-10D
BD Medical
No.:309624
Capacity: 1mL; 21 guage needle
Pack of 100 for $26.07
Case of 8 PK for $160.31
N/A N/A
Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 
14-826-88
BD Medical
No.:309626
Capacity: 1cc/16minim; 25 guage needle
Pack of 100 for $25.72
Case of 8 PK for $158.51
N/A N/A
Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 
14-826-87
BD Medical
No.:309623
Capacity: 1mL; 27 guage needle
Pack of 100 for $26.07
Case of 8 PK for $160.41
N/A N/A
Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 
14-823-2E
BD Medical
No.:309625
Capacity: 1mL(intradermal); 26 guage needle
Pack of 100 for $36.90
Case of 8 PK for $185.73
N/A N/A
Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 
148232F
BD Medical
No.:309602
Capacity: 1mL; Without needle
Accessories
N/A N/A N/A
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.  Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 
Description
  • Color-coded needle package and hub In Easy Peel™ package; autoclavable out of package Available with or without detachable PrecisionGlide™ needle
  • Syringe 14-823-2F accepts PrecisionGlide, Yale™, and Perfektum™ disposable and reusable needles