Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Print Email
74-011-50
Thermo Scientific™ Shandon™ Cytoblock™ Cell Block Preparation System
Increases ability to process scanty or unseen material for cell block and reduces loss of critical material
  • Increases ability to process scanty or unseen material for cell block and reduces loss of critical material
  • Suitable for immunohistochemical techniques
  • Replaces time-consuming and costly agar and thrombin techniques
  • Requires phosphate-free fixative on processor, such as Thermo Scientific Shandon fixatives Zinc Formal-Fixx™, Formal-Fixx, or Glyo-Fixx™ (formalin substitute) or simply bypass formalin fixation steps on processor
  • Cytoblock system can also be run on a rapid biopsy program
View Specifications
Images for Thermo Scientific™ Shandon™ Cytoblock™ Cell Block Preparation System
  • 74-011-50
74-011-50
74-011-50
(2 item(s) in product Group)
  • Items
  • Specifications
Catalog No. Description Price per Unit Quantity & Availability Add Item(s)
74-011-50
Thermo Scientific
No.:7401150
Cell preparation system
Each for $116.45
Check Availability
Add
to Cart
(0) in My Cart
NC9614386
Thermo Scientific
No.:7401151
Replacement reagent
Each for $63.00
Check Availability
Add
to Cart
(0) in My Cart
Description
  • Increases ability to process scanty or unseen material for cell block and reduces loss of critical material
  • Suitable for immunohistochemical techniques
  • Replaces time-consuming and costly agar and thrombin techniques
  • Requires phosphate-free fixative on processor, such as Thermo Scientific Shandon fixatives Zinc Formal-Fixx™, Formal-Fixx, or Glyo-Fixx™ (formalin substitute) or simply bypass formalin fixation steps on processor
  • Cytoblock system can also be run on a rapid biopsy program