Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Print Email
JPL430A
View Image Gallery (1) Image(s) are representative of the product Group
  • JPL430A
Available on GSA/VA Contract for Federal Government customers only. Thermo Scientific* Jewett* High-Performance Plasma Freezers
Designed to meet the highest standards for sensitive plasma freezing
  • High-density, CFC-free, blown-in insulation conforms to cabinet shape, to reduce gaps and increase temperature uniformity
  • Powerful, industrial-grade compressors for optimal product protection
  • 1 in. insulated access port standard
  • Positive, forced-air circulation maintains temperature uniformity throughout the cabinet
  • Set of four 2 in. casters: two locking and two regular
  • Adjustable, stainless-steel drawers
View Specifications
Alert: Products on this page may include voltages not considered standard in North America. Please review carefully.
Images for Available on GSA/VA Contract for Federal Government customers only. Thermo Scientific* Jewett* High-Performance Plasma Freezers
  • JPL430A
JPL430A
(7 item(s) in product Group)
  • Items
  • Specifications
Description Catalog No. Availability Quantity Price
4.7 cu. ft. (133L); 115V 60Hz
JPL430A Available on GSA/VA Contract for Federal Government customers only. 
Thermo Scientific
No.:JPL430A
N/AYou must be logged in and have a valid account number to view item availability   Each for $6,666.00
(0) in My Cart
11.5 cu. ft (326L); 115V 60Hz
JPL1230A Available on GSA/VA Contract for Federal Government customers only. 
Thermo Scientific
No.:JPL1230A
N/AYou must be logged in and have a valid account number to view item availability   Each for $9,054.00
(0) in My Cart
23.3 cu. ft. (659L); 115V 60Hz
JPL2330A Available on GSA/VA Contract for Federal Government customers only. 
Thermo Scientific
No.:JPL2330A
N/AYou must be logged in and have a valid account number to view item availability   Each for $10,932.00
(0) in My Cart
29.2 cu. ft. (826L); 115V 60Hz
JPL3030A Available on GSA/VA Contract for Federal Government customers only. 
Thermo Scientific
No.:JPL3030A
N/AYou must be logged in and have a valid account number to view item availability   Each for $12,205.00
(0) in My Cart
29.2 cu. ft. (826L); Holds 665 boxes; 1 door, 7 drawers; 208/240V, 60Hz
JPL3030D Available on GSA/VA Contract for Federal Government customers only. 
Thermo Scientific
No.:JPL3030D
N/AYou must be logged in and have a valid account number to view item availability   Each for $12,205.00
(0) in My Cart
51.1 cu. ft. (1447L); 115V 60Hz
JPL5030A Available on GSA/VA Contract for Federal Government customers only. 
Thermo Scientific
No.:JPL5030A
N/AYou must be logged in and have a valid account number to view item availability   Each for $14,815.00
(0) in My Cart
51.1 cu. ft. (1447L); 208/240V 60Hz
JPL5030D Available on GSA/VA Contract for Federal Government customers only. 
Thermo Scientific
No.:JPL5030D
N/AYou must be logged in and have a valid account number to view item availability   Each for $14,815.00
(0) in My Cart
Add Items to Cart
Description
  • High-density, CFC-free, blown-in insulation conforms to cabinet shape, to reduce gaps and increase temperature uniformity
  • Powerful, industrial-grade compressors for optimal product protection
  • 1 in. insulated access port standard
  • Positive, forced-air circulation maintains temperature uniformity throughout the cabinet
  • Set of four 2 in. casters: two locking and two regular
  • Adjustable, stainless-steel drawers
  • Smooth, ergonomic handle
  • Lockable doors for added sample security
  • Standard inkless, seven-day graphic temperature chart recorder
  • Bright digital temperature display
  • Microprocessor control system with audio and visual alarms
  • Preset -30°C cabinets
  • Hospital-grade plug
Certifications: UL, CE
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your 9-digit account number, or become a registered user on fishersci.com by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service at 1-800-766-7000.