Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Print Email
22-286-239
Pro-Lab Diagnostics Vision™ Polyvalent Shigella Slide Agglutination Antisera
Intended to aid in the serological identification of Shigella organisms
  • Prepared in rabbits using reference strains according to recognized guidelines and absorbed to remove cross-reactions
  • Supplied in 2.0mL dropper bottles in a ready-to-use format
View Specifications
Images for Pro-Lab Diagnostics Vision™ Polyvalent Shigella Slide Agglutination Antisera
  • 22-286-239
22-286-239
22-286-239
(4 item(s) in product Group)
  • Items
  • Specifications
Catalog No. Description Price per Unit Quantity & Availability Add Item(s)
22-286-238
Pro-Lab Diagnostics
No.:PL.6900
Shigella sonnei Phase 1 and 2
22-286-239
Pro-Lab Diagnostics
No.:PL.6901
Shigella flexneri 1-6 X and Y
22-286-240
Pro-Lab Diagnostics
No.:PL.6902
Shigella dysenteriae 1-10
22-286-241
Pro-Lab Diagnostics
No.:PL.6903
Shigella boydii 1-15
Description
  • Prepared in rabbits using reference strains according to recognized guidelines and absorbed to remove cross-reactions
  • Supplied in 2.0mL dropper bottles in a ready-to-use format