Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Print Email
AB-0757 (shown with plate, sold separately)
Thermo Scientific™ Easy Pierce Heat Sealing Foil
Ideal for compound management and high-throughput screening applications
  • Suitable for PCR (requires the pressure from a screw-down heated lid)
  • Easily pierced
  • Peelable bond can be removed by hand
  • 20µm foil is even more easily pierced and is suitable for multiple layering
  • Recommended for use with DMSO
  • Seal Integrity Range: -80°C to +80°C (+120°C with adequate pressure from cycler lid)
View Specifications
Images for Thermo Scientific™ Easy Pierce Heat Sealing Foil
  • AB-0757 (shown with plate, sold separately)
AB-0757 (shown with plate, sold separately)
AB-0757 (shown with plate, sold separately)
(4 item(s) in product Group)
  • Items
  • Specifications
Catalog No. Description Price per Unit Quantity & Availability Add Item(s)
AB-0757
Thermo Scientific
No.:AB-0757
For ALPS 25 Automated Laboratory Plate Sealer; 38 microns; Sheets
Case of 100 for $95.71
Check Availability
Add
to Cart
(0) in My Cart
AB-3738
Thermo Scientific
No.:AB3738
For ALPS 300; 38 microns; Roll
Each for $1,624.84
Check Availability
Add
to Cart
(0) in My Cart
AB-3720
Thermo Scientific
No.:AB-3720
For ALPS 300; 20 microns, Roll
Each for $1,650.05
Check Availability
Add
to Cart
(0) in My Cart
AB-1720
Thermo Scientific
No.:AB1720
For ALPS 25 Automated Laboratory Plate Sealer; 20 microns, Sheets
Pack of 100 for $95.71
Check Availability
Add
to Cart
(0) in My Cart
Add Items to Cart
Description
  • Suitable for PCR (requires the pressure from a screw-down heated lid)
  • Easily pierced
  • Peelable bond can be removed by hand
  • 20µm foil is even more easily pierced and is suitable for multiple layering
  • Recommended for use with DMSO
  • Seal Integrity Range: -80°C to +80°C (+120°C with adequate pressure from cycler lid)