Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Print Email
View Image Gallery (2) Image(s) are representative of the product Group
Available on GSA/VA Contract for Federal Government customers only. Thermo Scientific™ Orion™ ISE Ionic Strength Adjustors (ISA) and Special Reagents
Ionic strength adjuster is added to all samples and standards to create a uniform background ionic strength, which provides more reproducible measurements.
View Specifications
Images for Available on GSA/VA Contract for Federal Government customers only. Thermo Scientific™ Orion™ ISE Ionic Strength Adjustors (ISA) and Special Reagents
(14 item(s) in product Group)
  • Items
  • Specifications
Catalog No. Description Price per Unit Quantity & Availability Add Item(s)
13-641-883 Available on GSA/VA Contract for Federal Government customers only. 
No.:951211
pH Adjusting ISA for Ammonia Electrode, 475mL
Each for $121.36
Check Availability
Add
to Cart
(0) in My Cart
13-641-852 Available on GSA/VA Contract for Federal Government customers only. 
No.:940011
ISA for Bromide, Cadmium, Chloride, Cupric, Iodide, Sufide, Thiocyanate Electrodes, 475mL
Each for $98.93
Check Availability
Add
to Cart
(0) in My Cart
13-641-770 Available on GSA/VA Contract for Federal Government customers only. 
No.:950210
ISA for Carbon Dioxide Electrode, 475mL
Each for $116.86
Check Availability
Add
to Cart
(0) in My Cart
13-641-954Available on GSA/VA Contract for Federal Government customers only. 
No.:934610
ISA for Nitrite Electrode, 475mL
MSDS
Each for $101.24
Check Availability
Add
to Cart
(0) in My Cart
13-641-748 Available on GSA/VA Contract for Federal Government customers only. 
No.:841111
ISA for Sodium Electrode, 475mL
Each for $67.88
Check Availability
Add
to Cart
(0) in My Cart
NC9975931
Thermo Scientific
No.:951213
Storage Solution for Ammonia Electrode, 475mL
Each for $70.68
Check Availability
Add
to Cart
(0) in My Cart
13-641-878 Available on GSA/VA Contract for Federal Government customers only. 
No.:941709
Complextion Ionic Strength Adjuster (CISA) for Chloride Electrode, 2 x 1L
Each for $312.02
Check Availability
Add
to Cart
(0) in My Cart
13-641-854 Available on GSA/VA Contract for Federal Government customers only. 
No.:977011
Acid Reagent for Residual Chlorine Electrode, 475mL
Each for $97.09
Check Availability
Add
to Cart
(0) in My Cart
13-641-823 Available on GSA/VA Contract for Federal Government customers only. 
No.:977010
Iodide Reagent for Residual Chlorine Electrode, 5 x 50mL
MSDS
Each for $112.51
Check Availability
Add
to Cart
(0) in My Cart
13-641-853 Available on GSA/VA Contract for Federal Government customers only. 
Thermo Scientific
No.:951011
Alkaline Reagent for Cyanide Electrode, 475mL
Each for $119.74
Check Availability
Add
to Cart
(0) in My Cart
13-642-578 Available on GSA/VA Contract for Federal Government customers only. 
No.:940909
TISAB II for Fluoride Electrode, 1 Gallon (3.8L)
MSDS
Each for $151.86
Check Availability
Add
to Cart
(0) in My Cart
13-641-873 Available on GSA/VA Contract for Federal Government customers only. 
No.:940999
TISAB II for Fluoride Electrode, 4 x 1 Gallon (4 x 3.8L)
MSDS
Case of 4 for $487.14
Check Availability
Add
to Cart
(0) in My Cart
13-641-874 Available on GSA/VA Contract for Federal Government customers only. 
No.:940911
TISAB III Concentrate for Fluoride Electrode, 475mL
Each for $178.48
Check Availability
Add
to Cart
(0) in My Cart
13-641-802 Available on GSA/VA Contract for Federal Government customers only. 
No.:930710
Nitrate Interference Suppressor Solution (NISS) for Nitrate Electrode, 475mL
Each for $99.58
Check Availability
Add
to Cart
(0) in My Cart
Add Items to Cart
Description
Ionic strength adjuster is added to all samples and standards to create a uniform background ionic strength, which provides more reproducible measurements.
The ammonia pH-adjusting ISA (951211) and low level ammonia pH-adjusting ISA (951210) both include a pH-indicating blue dye for instant verification of correct pH in ammonia samples and standards with no pH measurement required. The ammonia pH-adjusting ISA (951211) prevents complexation of ammonia and inhibits formation of metal hydroxides, allowing determination of total ammonia in the sample and increasing the accuracy of results. The low level ammonia pH-adjusting ISA (951210) is ideal for ammonia samples with a concentration of 10ppm and below that do not contain metal interferences.
All ionic strength adjusters and special reagents are packaged as one 1 pint (475mL) bottle, except:
  • Cat. No. 941709 is packaged as two 1 quart (2 x 1L) bottles
  • Cat. No. 941609 is packaged as four 1 pint (4 x 475mL) bottles
  • Cat. No. 940909 is packaged as one 1 gallon (3.7L) bottle
  • Cat. No. 940999 is packaged as four 1 gallon (4 x 3.7L) bottles
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your 9-digit account number, or become a registered user on fishersci.com by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service at 1-800-766-7000.