Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Print Email
13-636-AB15PA
View Image Gallery (2) Image(s) are representative of the product Group
  • 13-636-AB15PA
  • 13-636-AB15PA
Available on GSA/VA Contract for Federal Government customers only. Fisher Scientific™ accumet™ AB15+ Basic and BioBasic™ pH/mV/°C Meters
Compact, affordable pH/mV meters with intuitive operation
Includes: Meter Only package includes meter, power supply, electrode support (13-637-671) and operation manual.
Meter Kit package includes items above plus an accumet liquid-filled, glass body single-junction combination pH Ag/AgCl Electrode (13-620-285) and separate ATC probe (13-620-19).
BioBasic Meter Kit package for the life science lab includes meter, power supply, manual, electrode support and accuTupH* rugged bulb, liquid-filled, glass body double-junction combination pH Combination Electrode (13-620-183) and separate ATC probe (13-620-19).
Note: Each package is also available with a Certificate of Traceability to NIST™ Standards.
  • Intuitive operation and improved user interface Large LCD guides user with plain-language prompts and error messages
  • Display indicators keep you apprised of menu choice, measuring units, buffers standardized (in pH mode) and temperature
  • No cryptic icons to learn: prompts and status/error messages are displayed in plain language
  • Store up to 100 datapoints in nonvolatile memory
  • Selectable resolution to 0.1, 0.01 or 0.001pH
  • Five-button, soft-key input simplifies data entry
  • Internal diagnostics
  • Automatic recognition of 15 buffers and five-point calibration
  • BNC and reference pin inputs, input for accuFet™ Electrode, ATC probe input, plus bidirectional RS-232 port
View Specifications
Images for Available on GSA/VA Contract for Federal Government customers only. Fisher Scientific™ accumet™ AB15+ Basic and BioBasic™ pH/mV/°C Meters
  • 13-636-AB15PA
  • 13-636-AB15PA
13-636-AB15PA
13-636-AB15PA
(4 item(s) in product Group)
  • Items
  • Specifications
Catalog No. Description Price per Unit Quantity & Availability Add Item(s)
13-636-AB15PC Meter Kit with NIST-traceable certificate
N/A N/A N/A
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
13-636-AB15PA Meter only
Accessories
N/A N/A N/A
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
13-636-AB15PB BioBasic meter kit
Accessories
N/A N/A N/A
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
13-636-AB15P Meter kit
Accessories
N/A N/A N/A
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Add Items to Cart
Description
  • Intuitive operation and improved user interface Large LCD guides user with plain-language prompts and error messages
  • Display indicators keep you apprised of menu choice, measuring units, buffers standardized (in pH mode) and temperature
  • No cryptic icons to learn: prompts and status/error messages are displayed in plain language
  • Store up to 100 datapoints in nonvolatile memory
  • Selectable resolution to 0.1, 0.01 or 0.001pH
  • Five-button, soft-key input simplifies data entry
  • Internal diagnostics
  • Automatic recognition of 15 buffers and five-point calibration
  • BNC and reference pin inputs, input for accuFet™ Electrode, ATC probe input, plus bidirectional RS-232 port
Specifications & Ordering Information:
Dimensions: 7.5L x 5.5W x 3.25 in.H (19 x 14 x 8cm). Net weight: 1.875 lb. (0.8kg).
Warranty: accumet Model AB15+ is warranted to the end user to be free from defects in materials and workmanship under normal use for a period of three years.
Specifications
ModelAB15+
pH Mode
Range-1.999 to +19.999pH
Resolution0.1/0.01/0.001pH
Relative Accuracy±0.002pH ± 1 least significant digit
mV Mode
Range0 to ±1800mV (abs. or rel.)
Resolution0.1mV
Relative Accuracy±0.2mV
Temperature Mode
Range-5.0° to +105.0°C (23° to 221°F)
Resolution0.1°C
Relative Accuracy±0.2°C
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your 9-digit account number, or become a registered user on fishersci.com by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service at 1-800-766-7000.