Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Print Email
Evolution* 60S w/printer
Thermo Scientific™ Evolution™ 60S UV-Visible Spectrophotometer
From the technician to the research scientist, the Evolution 60S offers the ideal solution delivering power and flexibility in a compact package
Efficient optical design delivers a new level of clarity to your measurements.
Maximum Performance, Compact Design
  • Acquire accurate spectra with scan speeds up to 4200nm/min.
  • Depend on dual-beam optics for superior photometric accuracy during long measurements
  • Acquire measurements from the UV to the near-IR
  • Choose either a local control instrument or a computer-controlled model to match the needs of your laboratory
  • Increase sample throughput with the integrated 6-cell changer; save data to a USB memory device
  • Rely on accurate data with pre-programmed performance verification methods; save bench space with small footprint
  • Go beyond the Evolution 60S instrument with accessories available for every type of sample, Customize to fit your laboratory needs
View Specifications
Images for Thermo Scientific™ Evolution™ 60S UV-Visible Spectrophotometer
  • Evolution* 60S w/printer
Evolution* 60S w/printer
Evolution* 60S w/printer
(3 item(s) in product Group)
  • Items
  • Specifications
Catalog No. Description Price per Unit Quantity & Availability Add Item(s)
14-386-510
Thermo Scientific
No.:840-208500
Local control
Accessories
Each for $6,175.57
Check Availability
Add
to Cart
(0) in My Cart
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
14-386-511
Thermo Scientific
No.:840-208600
Local control w/printer
Accessories
Each for $6,675.97
Check Availability
Add
to Cart
(0) in My Cart
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
14-386-512
Thermo Scientific
No.:840-208700
PC control
Accessories
Each for $5,922.82
Check Availability
Add
to Cart
(0) in My Cart
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
Add Items to Cart
Description
Maximum Performance, Compact Design
  • Acquire accurate spectra with scan speeds up to 4200nm/min.
  • Depend on dual-beam optics for superior photometric accuracy during long measurements
  • Acquire measurements from the UV to the near-IR
  • Choose either a local control instrument or a computer-controlled model to match the needs of your laboratory
  • Increase sample throughput with the integrated 6-cell changer; save data to a USB memory device
  • Rely on accurate data with pre-programmed performance verification methods; save bench space with small footprint
  • Go beyond the Evolution 60S instrument with accessories available for every type of sample, Customize to fit your laboratory needs
Maintenance-Free Lamp
  • Save time with the instant-on xenon flash lamp
  • Perform accurate analysis over the entire wavelength range of 190-1100nm
  • Prevent damage to sensitive samples – does not continuously expose sample to intense UV light
  • Long lifetime xenon flash lamp – guaranteed for three years of continuous use
  • Xenon lamp produces no heat – sample compartment temperature does not change
Specifications
Optical Design Dual Beam – Internal Reference Detector
Spectral Bandwidth1.0nm
Light Source (Typical Lifetime)Xenon Flash Lamp (5 years typical; 3 years
guaranteed)
DetectorsDual Silicon Photodiodes
Wavelength Range190-1100nm
Photometric Linear Range>3.5 A
Stray Light<0.08%T at 220, 340nm (NaI, NaNO2) <1.0%T 198-200nm (KCl)
Resolution (Toluene in Hexane)>1.5
DisplayGraphical with LCD backlight 3.8 x 2.8 in. (9.7 x 7.1cm)
KeypadSealed Membrane with tactile response keys
Printer (optional)40 column Internal (text and graphics) External
USB printer (HP PCL 3.0 and greater)
ConnectivityUSB
Dimensions11.8W x 15.7D x 9.8 in.H (30 x 40 x 25cm)
Weight19 lb. (8.6kg)
Power RequirementsSelected Automatically 100–240V; 50/60Hz