Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Print Email
6751E
View Image Gallery (4) Image(s) are representative of the product Group
  • 6751E
  • 6751C20
  • 6751C4
  • 6751P
Available on GSA/VA Contract for Federal Government customers only. SPEX SamplePrep™ 6770 Freezer/Mill™ Accessories: Vial Set
For use with Spex SamplePrep Freezer/Mill Cryogenic Grinder
Vial Set 6751
  • Sample capacity: 0.5 to 4.0mL
  • Makes one complete vial plus three spare center sections
  • Includes stainless-steel impactor, two end plugs, and four polycarbonate center cylinders (6751C4)
  • Individual components available
View Specifications
Images for Available on GSA/VA Contract for Federal Government customers only. SPEX SamplePrep™ 6770 Freezer/Mill™ Accessories: Vial Set
  • 6751E
  • 6751C20
  • 6751C4
  • 6751P
6751E
6751E
(9 item(s) in product Group)
  • Items
  • Specifications
Catalog No. Description Price per Unit Quantity & Availability Add Item(s)
6751 Available on GSA/VA Contract for Federal Government customers only. 
SPEX CertiPrep
No.:6751
Grinding vial set
Each for $345.00
Check Availability
Add
to Cart
(0) in My Cart
6751C4 Available on GSA/VA Contract for Federal Government customers only. 
SPEX CertiPrep
No.:6751C4
Polycarbonate center cylinders, 4-pack
Pack of 4 for $129.30
Check Availability
Add
to Cart
(0) in My Cart
6751C20 Available on GSA/VA Contract for Federal Government customers only. 
SPEX CertiPrep
No.:6751C20
Polycarbonate center cylinders, 20-pack
Pack of 20 for $526.00
Check Availability
Add
to Cart
(0) in My Cart
6751EAvailable on GSA/VA Contract for Federal Government customers only. 
SPEX CertiPrep
No.:6751E
End plugs
Pair for $223.00
Check Availability
Add
to Cart
(0) in My Cart
6751P Available on GSA/VA Contract for Federal Government customers only. 
SPEX CertiPrep
No.:6751P
Impactor
Each for $96.70
Check Availability
Add
to Cart
(0) in My Cart
6752 Available on GSA/VA Contract for Federal Government customers only. 
SPEX CertiPrep
No.:6752
Steel center cylinder
Each for $193.40
Check Availability
Add
to Cart
(0) in My Cart
6754 Available on GSA/VA Contract for Federal Government customers only. 
SPEX CertiPrep
No.:6754
Extractor and vial opener set
Each for $525.00
Check Availability
Add
to Cart
(0) in My Cart
6755 Available on GSA/VA Contract for Federal Government customers only. 
SPEX CertiPrep
No.:6755
Vial rack
Each for $60.30
Check Availability
Add
to Cart
(0) in My Cart
6814-115
SPEX CertiPrep
No.:6814-115
Autoextractor, 115V/60hz
Each for $1,392.00
Check Availability
Add
to Cart
(0) in My Cart
Add Items to Cart
Description
Vial Set 6751
  • Sample capacity: 0.5 to 4.0mL
  • Makes one complete vial plus three spare center sections
  • Includes stainless-steel impactor, two end plugs, and four polycarbonate center cylinders (6751C4)
  • Individual components available
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your 9-digit account number, or become a registered user on fishersci.com by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service at 1-800-766-7000.