Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Print Email
17-720-414
View Image Gallery (6) Image(s) are representative of the product Group
  • 17-720-414
  • 17-720-414
  • 17-720-414
  • 17-720-414
  • 17-720-414
  • 17-720-414
Thermo Scientific™ FirstDefender™ RMX Handheld Chemical Identification
Designed for flexible use modes; uses Raman spectroscopy to quickly identify unknown solid and liquid chemicals down range
  • Sophisticated chemometric algorithms automatically determine presence of mixed and contaminated chemicals
  • Flexible modes—offers handheld use with fixed probe or vial, or easily connects to a tactical robot through integrated RS232 port for universal control
  • Designed for use down range with a tough form factor, quick decontamination and easy operation in bulky protective gear
  • Lightweight, ergonomically designed unit weighs 2 lb. (919g) for easy transportation down range
  • Point-and-shoot sampling offers improved operation through sealed translucent containers; avoids exposure and maintains evidence
  • Noncontact analysis requires no consumables and no calibration
  • The easy-to-navigate vast substance library includes explosives, toxic industrial chemicals (TICs), chemical warfare agents (CWAs), narcotics, precursors, white powders and more
  • Easy to use and maintain with a menu driven user interface that requires minimal training and no scheduled maintenance or calibration
  • Can be powered using the included rechargeable battery, wall adapter or SureFire™ disposable batteries
View Specifications
Images for Thermo Scientific™ FirstDefender™ RMX Handheld Chemical Identification
  • 17-720-414
  • 17-720-414
  • 17-720-414
  • 17-720-414
  • 17-720-414
  • 17-720-414
17-720-414
17-720-414
(5 item(s) in product Group)
  • Items
  • Specifications
Catalog No. Description Price per Unit Quantity & Availability Add Item(s)
17-720-414 Available on GSA/VA Contract for Federal Government customers only. 
THERMO SCI WILMINGTON
No.:800-00211-01
1-year warranty and support
N/A N/A N/A
This product is not available for sales online. Please call Customer Service at 1-800-766-7000 or send an email to cs.webemail@thermofisher.com for assistance. 
17-720-415 Available on GSA/VA Contract for Federal Government customers only. 
THERMO SCI WILMINGTON
No.:800-00212-01
2-year warranty and support
N/A N/A N/A
This product is not available for sales online. Please call Customer Service at 1-800-766-7000 or send an email to cs.webemail@thermofisher.com for assistance. 
17-720-416 Available on GSA/VA Contract for Federal Government customers only. 
THERMO SCI WILMINGTON
No.:800-00213-01
3-year warranty and support
N/A N/A N/A
This product is not available for sales online. Please call Customer Service at 1-800-766-7000 or send an email to cs.webemail@thermofisher.com for assistance. 
17-720-417 Available on GSA/VA Contract for Federal Government customers only. 
THERMO SCI WILMINGTON
No.:800-00214-01
4-year warranty and support
N/A N/A N/A
This product is not available for sales online. Please call Customer Service at 1-800-766-7000 or send an email to cs.webemail@thermofisher.com for assistance. 
17-720-418 Available on GSA/VA Contract for Federal Government customers only. 
THERMO SCI WILMINGTON
No.:800-00215-01
5-year warranty and support
N/A N/A N/A
This product is not available for sales online. Please call Customer Service at 1-800-766-7000 or send an email to cs.webemail@thermofisher.com for assistance. 
Description
  • Sophisticated chemometric algorithms automatically determine presence of mixed and contaminated chemicals
  • Flexible modes—offers handheld use with fixed probe or vial, or easily connects to a tactical robot through integrated RS232 port for universal control
  • Designed for use down range with a tough form factor, quick decontamination and easy operation in bulky protective gear
  • Lightweight, ergonomically designed unit weighs 2 lb. (919g) for easy transportation down range
  • Point-and-shoot sampling offers improved operation through sealed translucent containers; avoids exposure and maintains evidence
  • Noncontact analysis requires no consumables and no calibration
  • The easy-to-navigate vast substance library includes explosives, toxic industrial chemicals (TICs), chemical warfare agents (CWAs), narcotics, precursors, white powders and more
  • Easy to use and maintain with a menu driven user interface that requires minimal training and no scheduled maintenance or calibration
  • Can be powered using the included rechargeable battery, wall adapter or SureFire™ disposable batteries
Certifications: MIL-STD 810G and IP67 certified.
Specifications
Raman Spectral Range250cm-1 to 2875cm-1
Spectral Resolution7 to 10.5cm-1 (FWHM) across range
Laser (excitation wavelength)785nm ±0.5nm, 2cm-1 linewidth, stability <0.1 cm-1
Laser Output PowerAdjustable, 75mW, 125mW, 250mW
Collection OpticsNA = 0.23, 17mm working distance; 0.14 to 1.8mm spot size
ExposureManual, Automatic modes (5ms min.)
BatteryRechargeable internal lithium ion battery or disposable batteries; >4 hours operation
Use ModeFlexible: handheld with fixed probe, vial mode, or robot-mounted
Operating Temperature-20°C to +40°C (continuous)
External Power SupplyDC Wall Adapter, 12V 1.25A
Robot IntegrationIntegration kit required; contact your customer service representative for more information
Size7.7 x 4.5 x 2.4 (19.6 x 11.4 x 6.1cm)
Weight2.0 lb. (919g)