Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Print Email
09-528-110B
View Image Gallery (3) Image(s) are representative of the product Group
  • 09-528-110B
  • 09-528-110B
Available on GSA/VA Contract for Federal Government customers only. Thermo Scientific™ Owl™ EasyCast™ B2 Mini Gel Electrophoresis Systems
Provide superior flexibility in a small footprint
The EasyCast systems feature leakproof casting that allows a gel to be cast and run in the same chamber with no tape or additional parts required. The EasyCast B2's longer gel length and the 2 comb slots provide the flexibility of running 8 to 48 samples on 1 gel. The optional multi load tray can increase the maximum number of samples to 108 in an 8 or 12 multichannel pipette format.
Includes: Buffer chamber, SuperSafe™ lid with attached power supply leads, EasyCast gasketed U.V. Transmissible (UVT) gel tray, 2 combs: 12 & 20 well, double-sided, 1.0/1.5 mm thick
  • 2 comb slots on the U.V. Transmissible (UVT) gel tray double sample capacity by doubling the number of sample wells
  • UVT gel trays are silk screened with a florescent ruler for easy measurement of bands
View Specifications
Images for Available on GSA/VA Contract for Federal Government customers only. Thermo Scientific™ Owl™ EasyCast™ B2 Mini Gel Electrophoresis Systems
  • 09-528-110B
  • 09-528-110B
09-528-110B
09-528-110B
(2 item(s) in product Group)
  • Items
  • Specifications
Catalog No. Description Price per Unit Quantity & Availability Add Item(s)
09-528-110B Available on GSA/VA Contract for Federal Government customers only. 
Thermo Scientific
No.:B2
EasyCast
Accessories
Each for $625.25
Check Availability
Add
to Cart
(0) in My Cart
09-528-118
Thermo Scientific
No.:B2-BP
EasyCast w/buffer exchange ports
Accessories
Each for $769.51
Check Availability
Add
to Cart
(0) in My Cart
Description
  • 2 comb slots on the U.V. Transmissible (UVT) gel tray double sample capacity by doubling the number of sample wells
  • UVT gel trays are silk screened with a florescent ruler for easy measurement of bands
Specifications & Ordering Information:
Available with optional buffer exchange ports (B2-BP) for use with an external recirculation pump
Certifications: CE marked
Warranty: Three years
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your 9-digit account number, or become a registered user on fishersci.com by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service at 1-800-766-7000.