Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Print Email
23-043-018
Available on GSA/VA Contract for Federal Government customers only. Quidel™ QuickVue™ Infectious Mononucleosis Test
Test for whole blood, serum or plasma, cassette with ± format
Includes: 20 Test cassettes, developer, mono negative control, mono positive control, 20 sample pipettes, 20 capillary tubes, package insert, procedure card
  • CLIA Complexity: Waived for whole blood
  • IgM-specific test
  • Results in five minutes
  • Easy to read ± test result
  • 99% sensitivity and 99% specificity
  • Flexible specimen collection options
View Specifications
Images for Available on GSA/VA Contract for Federal Government customers only. Quidel™ QuickVue™ Infectious Mononucleosis Test
  • 23-043-018
23-043-018
23-043-018
(1 item(s) in product Group)
  • Items
  • Specifications
Catalog No. Description Price per Unit Quantity & Availability Add Item(s)
23-043-018 Available on GSA/VA Contract for Federal Government customers only. 
Quidel
No.:20121
20 Tests
Pack of 20 for $205.16
Pack of 20 for $143.61 Online Instant Savings (30%)
Case of 12 PK for $2,215.94
Case of 12 PK for $1,551.16 Online Instant Savings (30%)
Check Availability
Add
to Cart
(0) in My Cart
Description
  • CLIA Complexity: Waived for whole blood
  • IgM-specific test
  • Results in five minutes
  • Easy to read ± test result
  • 99% sensitivity and 99% specificity
  • Flexible specimen collection options
Recommended Storage: Store at room temperature for up to 18 months from date of manufacture.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your 9-digit account number, or become a registered user on fishersci.com by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service at 1-800-766-7000.