Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Print Email
Normal Serum, Goat, 10mL
Thermo Scientific™ Pierce™ Normal Serum
Non-immune sera from goat, human, mouse, rabbit and other species
Normal serum that is derived from the same or related species of animal as a tissue sample or from the host species of specific antibodies used in immunodetection method provides several properties that improve assay performance for immunohistochemistry. Normal serum provides a sufficient quantity of endogenous proteins to saturate and block nonspecific binding sites on fixed samples or assay substrates. When used as an antibody diluent, non-immune serum also supplies a native chemical environment for antibodies to function properly without adding foreign protein components.
Highlights:
  • Multiple species available – choose normal serum from goat, horse, human, mouse, rabbit, rat and swine
  • Trouble-free formulation – delipidated (lipid-extracted) and dialyzed against weak phosphate-buffered saline (PBS) containing sodium azide
  • Stable format – supplied lyophilized (from PBS with preservative) for long shelf life and easy reconstitution in water
  • Safe source – USA is country of origin for all these animal-derived products
View Specifications
Images for Thermo Scientific™ Pierce™ Normal Serum
  • Normal Serum, Goat, 10mL
Normal Serum, Goat, 10mL
Normal Serum, Goat, 10mL
(10 item(s) in product Group)
  • Items
  • Specifications
Catalog No. Description Price per Unit Quantity & Availability Add Item(s)
PI-31872
Thermo Scientific
No.:31872
Goat Serum; 2mL
Pack for $36.93
Check Availability
Add
to Cart
(0) in My Cart
PI-31873
Thermo Scientific
No.:31873
Goat Serum; 10mL
Pack for $104.45
Check Availability
Add
to Cart
(0) in My Cart
PI-31874
Thermo Scientific
No.:31874
Horse Serum; 2mL
Pack for $36.93
Check Availability
Add
to Cart
(0) in My Cart
PI-31876
Thermo Scientific
No.:31876
Human Serum; 2mL
Pack for $36.93
Check Availability
Add
to Cart
(0) in My Cart
PI-31880
Thermo Scientific
No.:31880
Mouse Serum; 2mL
Pack for $47.48
Check Availability
Add
to Cart
(0) in My Cart
PI-31881
Thermo Scientific
No.:31881
Mouse Serum; 5mL
Pack for $68.58
Check Availability
Add
to Cart
(0) in My Cart
PI-31883
Thermo Scientific
No.:31883
Rabbit Serum; 5mL
Pack for $68.58
Check Availability
Add
to Cart
(0) in My Cart
PI-31884
Thermo Scientific
No.:31884
Rabbit Serum; 2mL
Pack for $36.93
Check Availability
Add
to Cart
(0) in My Cart
PI-31888
Thermo Scientific
No.:31888
Rat Serum; 2mL
Pack for $36.93
Check Availability
Add
to Cart
(0) in My Cart
PI-31890
Thermo Scientific
No.:31890
Swine Serum; 2mL
Each for $36.93
Check Availability
Add
to Cart
(0) in My Cart
Add Items to Cart
Description
Highlights:
  • Multiple species available – choose normal serum from goat, horse, human, mouse, rabbit, rat and swine
  • Trouble-free formulation – delipidated (lipid-extracted) and dialyzed against weak phosphate-buffered saline (PBS) containing sodium azide
  • Stable format – supplied lyophilized (from PBS with preservative) for long shelf life and easy reconstitution in water
  • Safe source – USA is country of origin for all these animal-derived products