Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Print Email
15-059-141, -142, -143
Ertco™ Exact-Temp™ Mercury Thermometers
For total immersion
Mounted in shatterproof plastic bottle filled with liquid to ensure accurate readings despite opening and closing of instrument door.
  • Certified traceable
  • With engraved stems and thoroughly annealed for accuracy
  • Thermometers are coated with PTFE resin to absorb shock and to contain glass and liquid in case of damage
  • With individual serial numbers
  • With double magnet for wall or door mounting
View Specifications
Alert: The products indicated contain mercury. Do not put products containing mercury in the trash. They should be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.
Images for Ertco™ Exact-Temp™ Mercury Thermometers
  • 15-059-141, -142, -143
15-059-141, -142, -143
15-059-141, -142, -143
(4 item(s) in product Group)
  • Items
  • Specifications
Catalog No. Description Price per Unit Quantity & Availability Add Item(s)
15059143
LUDWIG SCHNEIDER
No.:I-030-1
Type: Incubator; Range: 18° to 50° C
N/A N/A N/A
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
15059144
LUDWIG SCHNEIDER
No.:OV-070
Type: Oven; Range: 20° to 130° C
N/A N/A N/A
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
15059141
LUDWIG SCHNEIDER
No.:F-010-1
Type: Freezer; Range: - 30deg. to 0deg.C
N/A N/A N/A
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
15059142
LUDWIG SCHNEIDER
No.:R-020-1
Type: Refrigerator; Range: - 5° to + 15° C
N/A N/A N/A
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Add Items to Cart
Description
  • Certified traceable
  • With engraved stems and thoroughly annealed for accuracy
  • Thermometers are coated with PTFE resin to absorb shock and to contain glass and liquid in case of damage
  • With individual serial numbers
  • With double magnet for wall or door mounting
Certifications: A certificate is provided to indicate traceability to standards provided by NIST™