Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Print Email
Steri-Bottles Coliform Sampling
Thermo Scientific™ Screw-top Sterile Coliform Water Sample Bottle; Polystyrene, polyethylene, or polypropylene bottles with screw closure
Meets EPA Requirement for Bact-T sampling
Includes: Bottles assembled with caps
  • Polystyrene (PS) bottles are optically clear for easy sample viewing
  • Threaded screw closure for easy aseptic sample collection and secure cap engagement
  • Polyethylene (PE) and polypropylene (PP) bottles are contact clear allowing visibility of color changes while providing impact resistance for safer shipping and handling
  • Non-autofluorescent
  • Designed for total coliform and fecal coliform water sample analysis
  • Certified Sterile
  • Available pre-charged with a sodium thiosulfate (Na2S2O3) tablet for sample de-chlorination, or without a tablet for un-chlorinated water sources
  • 100 mL fill line
View Specifications
Images for Thermo Scientific™ Screw-top Sterile Coliform Water Sample Bottle; Polystyrene, polyethylene, or polypropylene bottles with screw closure
  • Steri-Bottles Coliform Sampling
Steri-Bottles Coliform Sampling
Steri-Bottles Coliform Sampling
(3 item(s) in product Group)
  • Items
  • Specifications
Catalog No. Description Price per Unit Quantity & Availability Add Item(s)
12-101-509
Thermo Scientific
No.:P156-120ST
120mL polystyrene bottle w/50mg tablet
Case of 200 for $414.50
Check Availability
Add
to Cart
(0) in My Cart
12-101-508
Thermo Scientific
No.:P156-120PP
120mL polypropylene bottle w/50mg tablet
Case of 200 for $356.00
Check Availability
Add
to Cart
(0) in My Cart
12-100-274
Thermo Scientific
No.:P156-150ST
150mL polystyrene bottle w/50mg tablet
Case of 345 for $837.00
Check Availability
Add
to Cart
(0) in My Cart
Add Items to Cart
Description
  • Polystyrene (PS) bottles are optically clear for easy sample viewing
  • Threaded screw closure for easy aseptic sample collection and secure cap engagement
  • Polyethylene (PE) and polypropylene (PP) bottles are contact clear allowing visibility of color changes while providing impact resistance for safer shipping and handling
  • Non-autofluorescent
  • Designed for total coliform and fecal coliform water sample analysis
  • Certified Sterile
  • Available pre-charged with a sodium thiosulfate (Na2S2O3) tablet for sample de-chlorination, or without a tablet for un-chlorinated water sources
  • 100 mL fill line
Applications: Coliform Bacteria sample collection and analysis
Compliance: Certified Sterile; manufactured under an ISO certified quality management system.
†Colilert is a registered trade mark of Idexx Laboratories, Inc.