Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Print Email
Filter Tip Reload family
View Image Gallery (2) Image(s) are representative of the product Group
  • Filter Tip Reload family
  • Single stacked rack
Fisherbrand™ SureOne™ Filter Tip Reload Pipet Tips
Sterile and free of RNase, DNase, ATP, pyrogen, bioburden, PCR inhibitors, endotoxin and DNA
Optimal
Packaging Features
  • Offset wells allow filter tips to stack
  • Rigid inserts are multi-channel format ensuring secure tip seal
  • Lot numbered inserts for traceability also include easy to read tip volume and Fisher catalog number
  • Sterile packaging with outer sleeve protects tips from contamination while refilling inserts
  • Environmentally friendly reload packaging reduces plastic waste and saves space
  • Blister sleeve guide rails align tip inserts for smooth and reliable transfer into the rack
Rack Features
  • Flush Fit Inserts sit flush with rack, allowing easy multichannel pipetting
  • Recycle Logo molded into all rack components
  • Notched clasp props lid open for autoclave venting
  • Sure Stack lid rim prevents racks from sliding around when stacked and carted around a lab
  • Safe-Lock latch allows for one-handed open and closing ability and won't pop open when dropped
  • Sure-snap design securely holds insert in place, preventing it from being lifted out while pipetting
  • Flip-lid can be either a hinged or lift-off lid; hinged format does not prevent use of a multichannel pipet
Insert Features
  • Inserts are easy to place onto and remove from rack; clip onto rack for a secure fit
  • Lot traceable—all inserts contain the Fisherbrand part number, tip volume, lot number
  • Indented insert holes inhibit tip movement, stabilizing them for multichannel work
  • Contact Clip helps prevent unnecessary contact with the tips while clipping in inserts
  • Multichannel design; all inserts are 96-well format
  • Robust non-pliable design provides secure seals on every tip
View Specifications
Images for Fisherbrand™ SureOne™ Filter Tip Reload Pipet Tips
  • Filter Tip Reload family
  • Single stacked rack
Filter Tip Reload family
Filter Tip Reload family
(9 item(s) in product Group)
  • Items
  • Specifications
Catalog No. Description Price per Unit Quantity & Availability Add Item(s)
02-707-472 Micropoint Nano tip; Graduated at 2.5uL; 0.1-5uL
Pack of 960 for $119.41
Pack of 960 for $77.62 Online Instant Savings (35%)
Case of 5 PK for $550.43
Case of 5 PK for $357.78 Online Instant Savings (35%)
Check Availability
Add
to Cart
(0) in My Cart
02-707-473 Micropoint tip; Graduated at 2uL; 0.1-10uL
Pack of 960 for $119.41
Pack of 960 for $77.62 Online Instant Savings (35%)
Case of 5 PK for $550.43
Case of 5 PK for $357.78 Online Instant Savings (35%)
Check Availability
Add
to Cart
(0) in My Cart
02-707-474 Low Retention Micropoint tip; Graduated at 2, 5, 10, 20uL; Extended length; 0.1-10uL
Pack of 960 for $119.41
Pack of 960 for $77.62 Online Instant Savings (35%)
Case of 5 PK for $550.43
Case of 5 PK for $357.78 Online Instant Savings (35%)
Check Availability
Add
to Cart
(0) in My Cart
02-707-475 Low retention micropoint tip; Graduated at 2, 5, 10, 20µL; 0.1 to 20uL
Pack of 960 for $119.41
Pack of 960 for $77.62 Online Instant Savings (35%)
Case of 5 PK for $550.43
Case of 5 PK for $357.78 Online Instant Savings (35%)
Check Availability
Add
to Cart
(0) in My Cart
02-707-476 Low Retention Beveled tip; Non graduated; 2-20uL
Pack of 960 for $119.62
Pack of 960 for $77.75 Online Instant Savings (35%)
Case of 5 PK for $537.10
Case of 5 PK for $349.12 Online Instant Savings (35%)
Check Availability
Add
to Cart
(0) in My Cart
02-707-477 Low Retention Beveled tip; Non graduated; 10-100uL
Pack of 960 for $119.41
Pack of 960 for $77.62 Online Instant Savings (35%)
Case of 5 PK for $550.43
Case of 5 PK for $357.78 Online Instant Savings (35%)
Check Availability
Add
to Cart
(0) in My Cart
02-707-478 Low Retention Micropoint tip; Graduated at 10, 50, 100, 200uL; 20-200uL
Pack of 960 for $119.41
Pack of 960 for $77.62 Online Instant Savings (35%)
Case of 5 PK for $550.43
Case of 5 PK for $357.78 Online Instant Savings (35%)
Check Availability
Add
to Cart
(0) in My Cart
02-707-479 Low Retention Micropoint tip; Graduated at 10, 50, 100, 200uL; Extended length; 30-300uL
Pack of 960 for $119.41
Pack of 960 for $77.62 Online Instant Savings (35%)
Case of 5 PK for $550.43
Case of 5 PK for $357.78 Online Instant Savings (35%)
Check Availability
Add
to Cart
(0) in My Cart
02-707-480 Micropoint tip; Graduated at 100, 200, 500, 1000uL; Extended length; 100-1000uL
Pack of 560 for $104.14
Pack of 560 for $67.69 Online Instant Savings (35%)
Case of 5 PK for $467.40
Case of 5 PK for $303.81 Online Instant Savings (35%)
Check Availability
Add
to Cart
(0) in My Cart
Add Items to Cart
Description
Packaging Features
  • Offset wells allow filter tips to stack
  • Rigid inserts are multi-channel format ensuring secure tip seal
  • Lot numbered inserts for traceability also include easy to read tip volume and Fisher catalog number
  • Sterile packaging with outer sleeve protects tips from contamination while refilling inserts
  • Environmentally friendly reload packaging reduces plastic waste and saves space
  • Blister sleeve guide rails align tip inserts for smooth and reliable transfer into the rack
Rack Features
  • Flush Fit Inserts sit flush with rack, allowing easy multichannel pipetting
  • Recycle Logo molded into all rack components
  • Notched clasp props lid open for autoclave venting
  • Sure Stack lid rim prevents racks from sliding around when stacked and carted around a lab
  • Safe-Lock latch allows for one-handed open and closing ability and won't pop open when dropped
  • Sure-snap design securely holds insert in place, preventing it from being lifted out while pipetting
  • Flip-lid can be either a hinged or lift-off lid; hinged format does not prevent use of a multichannel pipet
Insert Features
  • Inserts are easy to place onto and remove from rack; clip onto rack for a secure fit
  • Lot traceable—all inserts contain the Fisherbrand part number, tip volume, lot number
  • Indented insert holes inhibit tip movement, stabilizing them for multichannel work
  • Contact Clip helps prevent unnecessary contact with the tips while clipping in inserts
  • Multichannel design; all inserts are 96-well format
  • Robust non-pliable design provides secure seals on every tip