Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Print Email
Thermo Scientific MAS UA Controls
Available on GSA/VA Contract for Federal Government customers only. Thermo Scientific™ MAS™ UA Controls
Bi-level control designed to monitor urine dipstick and microscopic components
Includes: Includes 10 commonly tested dipstick tests plus microalbumin, creatinine and hCG
  • Liquid, ready to use
  • Human urine matrix
  • Assayed for most major reagent strips and strip systems
View Specifications
Images for Available on GSA/VA Contract for Federal Government customers only. Thermo Scientific™ MAS™ UA Controls
  • Thermo Scientific MAS UA Controls
Thermo Scientific MAS UA Controls
Thermo Scientific MAS UA Controls
(5 item(s) in product Group)
  • Items
  • Specifications
Catalog No. Description Price per Unit Quantity & Availability Add Item(s)
23-038-380 Available on GSA/VA Contract for Federal Government customers only. 
Thermo Scientific
No.:UAB115
Assayed; Level 1; Normal; 6 x 15mL
MSDS
N/A N/A N/A
This product cannot be ordered online due to special handling requirements. Please contact National Order Desk customer service at CS.NOD@thermofisher.com or 1-800-361-9402 for ordering assistance.  This product is regulated by the federal government, and may not be ordered online. 
23-038-381 Available on GSA/VA Contract for Federal Government customers only. 
Thermo Scientific
No.:UAB215
6 x 15mL
MSDS
N/A N/A N/A
This product cannot be ordered online due to special handling requirements. Please contact National Order Desk customer service at CS.NOD@thermofisher.com or 1-800-361-9402 for ordering assistance.  This product is regulated by the federal government, and may not be ordered online. 
23-038-384 Available on GSA/VA Contract for Federal Government customers only. 
Thermo Scientific
No.:UAB-160
Assayed; Level 1; Normal; 4 x 60mL
MSDS
N/A N/A N/A
This product cannot be ordered online due to special handling requirements. Please contact National Order Desk customer service at CS.NOD@thermofisher.com or 1-800-361-9402 for ordering assistance.  This product is regulated by the federal government, and may not be ordered online. 
23-038-385 Available on GSA/VA Contract for Federal Government customers only. 
Thermo Scientific
No.:UAB-260
4 x 60mL
MSDS
N/A N/A N/A
This product cannot be ordered online due to special handling requirements. Please contact National Order Desk customer service at CS.NOD@thermofisher.com or 1-800-361-9402 for ordering assistance.  This product is regulated by the federal government, and may not be ordered online. 
23-038-383 Available on GSA/VA Contract for Federal Government customers only. 
Thermo Scientific
No.:UABMP
Assayed; Bi-level; Multi-Pack; 6 x 15mL
MSDS
N/A N/A N/A
This product cannot be ordered online due to special handling requirements. Please contact National Order Desk customer service at CS.NOD@thermofisher.com or 1-800-361-9402 for ordering assistance.  This product is regulated by the federal government, and may not be ordered online. 
Description
  • Liquid, ready to use
  • Human urine matrix
  • Assayed for most major reagent strips and strip systems
Specifications & Ordering Information:
Available in convenient 15mL dropper bottles
Recommended Storage:
  • 24 month shelf life at 2° to 8°C
  • Open Vial: Refrigerated 2° to 8°C for 3 months
  • Room Temperature 18° to 25°C for 6 weeks
Analytes†
BilirubinNitrite
BloodpH
CreatininePotassium
CrystalsProtein
GlucoseRed Blood Cells
hCGSodium
KetonesSpecific Gravity
Leukocyte EsteraseUrobilinogen
MicroalbuminWhite Cells
† Refer to package insert for specific analyte and stability claims.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your 9-digit account number, or become a registered user on fishersci.com by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service at 1-800-766-7000.