Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Print Email
RGL2304
View Image Gallery (4) Image(s) are representative of the product Group
  • RGL2304
  • RGL2304
  • RGL3004 Series
  • RGL7504 Series
Available on GSA/VA Contract for Federal Government customers only. Thermo Scientific™ Revco™ High-Performance Laboratory Refrigerators with Glass Doors
Cabinet sizes to fit your space and storage needs
Revco high-performance lab refrigerators with glass doors are available in a wide range of cabinet sizes to fit virtually any application need. Each refrigerator comes with an advanced microprocessor temperature control, high-quality cabinet construction and advanced alarm systems. Postivie, forced-air circulation maintains temperature uniformity, protecting your valuable samples and inventory.
  • Adjustable temperatures between 1° and 8°C (factory preset at 4°C)
  • Automatic defrost and condensate removal
Microprocessor Control
  • Digital electronic technology designed to simplify operation and deliver greater accuracy
  • Icon-based controls
  • Large, easy-to-read digital display of temperature within 0.1°C
  • Graphic thermometer is steady green when temperature is normal, flashes top bar for overtemperature, flashes bulb for undertemperature
  • Sensor automatically initiates defrost cycle on demand, eliminating need for a defrost timer, minimizing frost buildup, and optimizing coil efficiency
Alarm System
  • Audible/visual warnings for temperature deviations or power failure
  • Key-operated ON/OFF/ALARM ON master switch for setpoint security
  • Push-button alarm test and remote alarm contacts
  • Alarm silence, ringback, and auto reset
  • Controller battery backup system
  • Alarm setpoints are preset to warn of temperature deviation at 5.5°C overtemperature and 1.5°C undertemperature
Construction
  • Positive forced-air circulation systems
  • Industrial-grade cabinet construction and compressors
  • High-density CFC-free urethane foam insulation
  • Self-closing, key-locked doors
  • Epoxy-coated, open-wire shelves adjustable in 1 in. (2.5cm) increments
  • Cold-rolled steel interior and exterior finished with high-impact powder paint coating
  • Interior lights with door-activated ON/OFF switch, independent switch on control panel, adjustable leveling feet (except 4.9 cu. ft. undercounter model)
  • 45.8 cu. ft. (1297L) units have double sliding glass doors, base-mounted controls and refrigeration systems, independent switch-activated lights, and dual-wheel 2 in. (5cm) casters instead of leveling feet
  • Undercounter model has a 1 in. (2.5cm) dia. access port with plug
View Specifications
Images for Available on GSA/VA Contract for Federal Government customers only. Thermo Scientific™ Revco™ High-Performance Laboratory Refrigerators with Glass Doors
  • RGL2304
  • RGL2304
  • RGL3004 Series
  • RGL7504 Series
RGL2304
RGL2304
(26 item(s) in product Group)
  • Items
  • Specifications
Catalog No. Description Price per Unit Quantity & Availability Add Item(s)
RGL404A Available on GSA/VA Contract for Federal Government customers only. 
Thermo Scientific
No.:RGL404A
4.9 cu. ft.; Glass door, 115V 60Hz
Each for $5,304.11
Check Availability
Add
to Cart
(0) in My Cart
RGL404D
Thermo Scientific
No.:RGL404D
4.9 cu. ft.; Glass door, NEMA 6-15P plug type; 208/230V 60Hz
Each for $5,314.33
Check Availability
Add
to Cart
(0) in My Cart
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
RGL404V
Thermo Scientific
No.:RGL404V
4.9 cu. ft.; Glass door, European plug type; 230V 50Hz
Each for $5,300.00
Check Availability
Add
to Cart
(0) in My Cart
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
RGL404W
Thermo Scientific
No.:RGL404W
4.9 cu. ft.; Glass door, U.K. plug type; 230V 50Hz
Each for $5,300.00
Check Availability
Add
to Cart
(0) in My Cart
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
RGL1204A Available on GSA/VA Contract for Federal Government customers only. 
Thermo Scientific
No.:RGL1204A
11.5 cu. ft.; Glass door, 115V 60Hz
Each for $5,754.00
Check Availability
Add
to Cart
(0) in My Cart
RGL1204D Available on GSA/VA Contract for Federal Government customers only. 
Thermo Scientific
No.:RGL1204D
11.5 cu. ft.; Glass door, NEMA 6-15P plug type; 208/230V 60Hz
Each for $5,774.44
Check Availability
Add
to Cart
(0) in My Cart
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
RGL1204V
Thermo Scientific
No.:RGL1204V
11.5 cu. ft.; Glass door, European plug type; 230V 50Hz
Each for $5,760.00
Check Availability
Add
to Cart
(0) in My Cart
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
RGL2304W
Thermo Scientific
No.:RGL2304W
23CF REF GLASS 230V23CF REF G
Each for $6,390.00
Check Availability
Add
to Cart
(0) in My Cart
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
RGL1204W
Thermo Scientific
No.:RGL1204W
11.5 cu. ft.; Glass door, U.K. plug type; 230V 50Hz
Each for $5,760.00
Check Availability
Add
to Cart
(0) in My Cart
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
RGL2304A Available on GSA/VA Contract for Federal Government customers only. 
Thermo Scientific
No.:RGL2304A
23.3 cu. ft.; Glass door, 115V 60Hz
Each for $6,387.13
Check Availability
Add
to Cart
(0) in My Cart
RGL2304D Available on GSA/VA Contract for Federal Government customers only. 
Thermo Scientific
No.:RGL2304D
23.3 cu. ft.; Glass door, NEMA 6-15P plug type; 208/230V, 60Hz
Each for $6,397.34
Check Availability
Add
to Cart
(0) in My Cart
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
RGL2304V
Thermo Scientific
No.:RGL2304V
23.3 cu. ft.; Glass door, European plug type; 230V 50Hz
Each for $6,390.00
Check Availability
Add
to Cart
(0) in My Cart
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
RGL3004A Available on GSA/VA Contract for Federal Government customers only. 
Thermo Scientific
No.:RGL3004A
29.9 cu. ft.; Glass door, 115V 60Hz
Each for $7,379.28
Check Availability
Add
to Cart
(0) in My Cart
RGL3004D Available on GSA/VA Contract for Federal Government customers only. 
Thermo Scientific
No.:RGL3004D
29.9 cu. ft.; Glass door, NEMA 6-15P plug type; 208/230V 60Hz
Each for $7,379.28
Check Availability
Add
to Cart
(0) in My Cart
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
RGL3004V
Thermo Scientific
No.:RGL3004V
29.9 cu. ft; Glass door, European plug type; 230V 50Hz
Each for $7,370.00
Check Availability
Add
to Cart
(0) in My Cart
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
RGL3004W
Thermo Scientific
No.:RGL3004W
29.9 cu. ft; Glass door, U.K. plug type; 230V 50Hz
Each for $7,370.00
Check Availability
Add
to Cart
(0) in My Cart
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
13-991-139 Available on GSA/VA Contract for Federal Government customers only. 
Thermo Scientific
No.:REL4504A
45.8 cu. ft.; 2 sliding glass doors, 115V 60Hz
Each for $8,441.29
Check Availability
Add
to Cart
(0) in My Cart
13-991-140 Available on GSA/VA Contract for Federal Government customers only. 
Thermo Scientific
No.:REL4504D
45.8 cu. ft.; 2 sliding glass doors, 230V 50Hz
Each for $8,441.29
Check Availability
Add
to Cart
(0) in My Cart
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
RGL5004A Available on GSA/VA Contract for Federal Government customers only. 
Thermo Scientific
No.:RGL5004A
51.1 cu. ft.; 2 glass doors, 115V 60Hz
Each for $9,576.03
Check Availability
Add
to Cart
(0) in My Cart
RGL5004D Available on GSA/VA Contract for Federal Government customers only. 
Thermo Scientific
No.:RGL5004D
51.1 cu. ft.; 2 glass doors, NEMA 6-15P plug type; 208/230V 60Hz
Each for $9,596.47
Check Availability
Add
to Cart
(0) in My Cart
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
RGL5004V
Thermo Scientific
No.:RGL5004V
51.1 cu. ft.; 2 glass doors, European plug type; 230V 50Hz
Each for $9,580.00
Check Availability
Add
to Cart
(0) in My Cart
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
RGL5004W
Thermo Scientific
No.:RGL5004W
51.1 cu.ft; Glass door, U.K. plug type; 230V 50Hz
Each for $9,580.00
Check Availability
Add
to Cart
(0) in My Cart
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
RGL7504A Available on GSA/VA Contract for Federal Government customers only. 
Thermo Scientific
No.:RGL7504A
78.8 cu. ft.; Glass door, 115V 60Hz
Each for $13,367.40
Check Availability
Add
to Cart
(0) in My Cart
RGL7504D Available on GSA/VA Contract for Federal Government customers only. 
Thermo Scientific
No.:RGL7504D
78.8 cu. ft.; Glass door, NEMA 6-15P plug type; 208/230V 60Hz
Each for $12,212.57
Check Availability
Add
to Cart
(0) in My Cart
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
RGL7504V
Thermo Scientific
No.:RGL7504V
78.8 cu. ft.; Glass door, European plug type; 230V 50Hz
Each for $12,190.00
Check Availability
Add
to Cart
(0) in My Cart
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
RGL7504W
Thermo Scientific
No.:RGL7504W
78.8 cu. ft.; Glass door, U.K. plug type; 230V 50Hz
Each for $12,190.00
Check Availability
Add
to Cart
(0) in My Cart
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
Add Items to Cart
Description
  • Adjustable temperatures between 1° and 8°C (factory preset at 4°C)
  • Automatic defrost and condensate removal
Microprocessor Control
  • Digital electronic technology designed to simplify operation and deliver greater accuracy
  • Icon-based controls
  • Large, easy-to-read digital display of temperature within 0.1°C
  • Graphic thermometer is steady green when temperature is normal, flashes top bar for overtemperature, flashes bulb for undertemperature
  • Sensor automatically initiates defrost cycle on demand, eliminating need for a defrost timer, minimizing frost buildup, and optimizing coil efficiency
Alarm System
  • Audible/visual warnings for temperature deviations or power failure
  • Key-operated ON/OFF/ALARM ON master switch for setpoint security
  • Push-button alarm test and remote alarm contacts
  • Alarm silence, ringback, and auto reset
  • Controller battery backup system
  • Alarm setpoints are preset to warn of temperature deviation at 5.5°C overtemperature and 1.5°C undertemperature
Construction
  • Positive forced-air circulation systems
  • Industrial-grade cabinet construction and compressors
  • High-density CFC-free urethane foam insulation
  • Self-closing, key-locked doors
  • Epoxy-coated, open-wire shelves adjustable in 1 in. (2.5cm) increments
  • Cold-rolled steel interior and exterior finished with high-impact powder paint coating
  • Interior lights with door-activated ON/OFF switch, independent switch on control panel, adjustable leveling feet (except 4.9 cu. ft. undercounter model)
  • 45.8 cu. ft. (1297L) units have double sliding glass doors, base-mounted controls and refrigeration systems, independent switch-activated lights, and dual-wheel 2 in. (5cm) casters instead of leveling feet
  • Undercounter model has a 1 in. (2.5cm) dia. access port with plug
Specifications & Ordering Information:
Alarm systems, temperature chart recorders, and power monitors are available separately. Other voltages are available; contact your Customer Service Representative for more information.
Compliance: UL and cUL listed. TUV approved. CE marked.
Warranty: Two years, parts and labor, US and Canada only
Specifications
Temperature Range1° to 8°C
Temperature ControlMicroprocessor
DisplayDigital
Temperature Resolution1°C
DefrostAutomatic
RefrigerantCFC- and HCFC-free R134A
CabinetScratch-resistant painted
ChamberScratch-resistant painted
InsulationHigh-density CFC-free urethane foam
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your 9-digit account number, or become a registered user on fishersci.com by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service at 1-800-766-7000.