Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Print Email
AB-0753
Thermo Scientific™ ABgene™ Thermo-Mat™ Sealing Mats
Round or square capped mats for sealing 96-well storage plates, available in autoclavable and piercable formats.
Thermo Scientific 96-well Sealing Mats are designed for 96-well storage plates and are available in either piercable or autoclavable formats. The round or square capped mats are made of thermoplastic elastomer and are suitable for long-term storage. Highlights Round or square capped mats for sealing 96-well storage plates · Alphanumeric grid-referencing· Molded from a proprietary thermoplastic elastomer for a flexible design· Suitable for long term storage· Available in autoclavable or piercable formats Pierceable version:· Solvent resistant (including DMSO, Ethanol, and Isopropanol)· Suitable for UV Irradiation and Gamma Irradiation· Requires 6N for piercing· Temperature Range: -20°C to +55°C Autoclavable version:· Suitable for UV Irradiation and Gamma Irradiation· Temperature Range: -20°C to +130°C
View Specifications
Images for Thermo Scientific™ ABgene™ Thermo-Mat™ Sealing Mats
  • AB-0753
AB-0753
AB-0753
(2 item(s) in product Group)
  • Items
  • Specifications
Catalog No. Description Price per Unit Quantity & Availability Add Item(s)
AB-0427 Available on GSA/VA Contract for Federal Government customers only. 
Thermo Scientific
No.:AB0427
For 96-Well Plate
Pack of 5 for $67.16
Check Availability
Add
to Cart
(0) in My Cart
AB-0770 Available on GSA/VA Contract for Federal Government customers only. 
Thermo Scientific
No.:AB-0770
For 384-Well Plate without Tabs
Pack of 5 for $134.20
Check Availability
Add
to Cart
(0) in My Cart
Description
Thermo Scientific 96-well Sealing Mats are designed for 96-well storage plates and are available in either piercable or autoclavable formats. The round or square capped mats are made of thermoplastic elastomer and are suitable for long-term storage. Highlights Round or square capped mats for sealing 96-well storage plates · Alphanumeric grid-referencing· Molded from a proprietary thermoplastic elastomer for a flexible design· Suitable for long term storage· Available in autoclavable or piercable formats Pierceable version:· Solvent resistant (including DMSO, Ethanol, and Isopropanol)· Suitable for UV Irradiation and Gamma Irradiation· Requires 6N for piercing· Temperature Range: -20°C to +55°C Autoclavable version:· Suitable for UV Irradiation and Gamma Irradiation· Temperature Range: -20°C to +130°C
Hazardous:NoStorage Condition:Ambient
Certifications: Certified free from DNase, RNase and human genomic DNA.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your 9-digit account number, or become a registered user on fishersci.com by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service at 1-800-766-7000.