Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Print Email
XX1104702
View Image Gallery (5) Image(s) are representative of the product Group
  • XX1104702
  • XX1104704
  • XX1104705
  • XX1104710
  • XX11J4750
Millipore* Sterifil* 47mm Aseptic Vacuum Filter System Replacement Parts
Used for general filtration applications and filtration of samples for particulate or biological contamination analysis
The closed unit protects sample and filtrate from environmental contamination.
The Sterifil holder and funnel available separately (without receiver flask and cover) to use with standard 1L vacuum filtering flask or multiple place manifold.
View Specifications
Images for Millipore* Sterifil* 47mm Aseptic Vacuum Filter System Replacement Parts
  • XX1104702
  • XX1104704
  • XX1104705
  • XX1104710
  • XX11J4750
XX1104702
(5 item(s) in product Group)
  • Items
  • Specifications
Description Catalog No. Availability Quantity Price
250mL Funnel
XX11 047 04
Millipore
No.:XX1104704
N/AYou must be logged in and have a valid account number to view item availability   Each for $23.85
(0) in My Cart
Receiver flask, 250mL
XX11 047 05
Millipore
No.:XX1104705
N/AYou must be logged in and have a valid account number to view item availability   Each for $30.49
(0) in My Cart
47mm Holder
XX11 047 10
Millipore
No.:XX1104710
N/AYou must be logged in and have a valid account number to view item availability   Each for $73.42
(0) in My Cart
With support screen
XX1104702
Millipore
No.:XX1104702
N/AYou must be logged in and have a valid account number to view item availability   Each for $10.53
(0) in My Cart
Sterifil Holder; 500mL
XX11J4750
Millipore
No.:XX11J4750
N/AYou must be logged in and have a valid account number to view item availability   Each for $126.73
(0) in My Cart
Add Items to Cart
Specifications & Ordering Information:
The Sterifil holder and funnel available separately (without receiver flask and cover) to use with standard 1L vacuum filtering flask or multiple place manifold.