Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Print Email
Available on GSA/VA Contract for Federal Government customers only. Fisherbrand™ SureOne Beveled Pipet Tips, Universal Fit: 1-200µL
Offers a full line of universal fit beveled pipet tips
  • All racked sterile tips are shrink wrapped and certified free of RNase, DNase, ATP, pyrogen, bioburden, PCR inhibitors, endotoxin, and DNA
  • All nonsterile racked and bulk nonfiltered tips are certified free of RNase, DNase, and DNA
View Specifications
Images for Available on GSA/VA Contract for Federal Government customers only. Fisherbrand™ SureOne Beveled Pipet Tips, Universal Fit: 1-200µL
(6 item(s) in product Group)
  • Items
  • Specifications
Catalog No. Description Price per Unit Quantity & Availability Add Item(s)
02-707-418 Available on GSA/VA Contract for Federal Government customers only.  Clear; Nonsterile; Hinged Rack, Packaging: HR, 10 x 96
Pack of 960 for $62.32
Pack of 960 for $43.62 Online Instant Savings (30%)
Case of 5 PK for $254.20
Case of 5 PK for $177.94 Online Instant Savings (30%)
Check Availability
Add
to Cart
(0) in My Cart
02-707-419 Available on GSA/VA Contract for Federal Government customers only.  Clear; Sterile; Hinged Rack; Packaging: HRS, 10 x 96
Pack of 960 for $66.83
Pack of 960 for $46.78 Online Instant Savings (30%)
Case of 5 PK for $282.90
Case of 5 PK for $198.03 Online Instant Savings (30%)
Check Availability
Add
to Cart
(0) in My Cart
02-707-420 Available on GSA/VA Contract for Federal Government customers only.  Clear; Nonsterile; Hinged Rack; Packaging: RD, 10 x 96
Pack of 960 for $48.79
Pack of 960 for $34.15 Online Instant Savings (30%)
Case of 5 PK for $208.07
Case of 5 PK for $145.65 Online Instant Savings (30%)
Check Availability
Add
to Cart
(0) in My Cart
02-707-421Available on GSA/VA Contract for Federal Government customers only.  Yellow; Nonsterile; Hinged Rack; Packaging: BP
Pack of 960 for $59.35
Pack of 960 for $41.55 Online Instant Savings (30%)
Case of 5 PK for $251.64
Case of 5 PK for $176.15 Online Instant Savings (30%)
Check Availability
Add
to Cart
(0) in My Cart
02-707-422 Available on GSA/VA Contract for Federal Government customers only.  Yellow; Sterile; Hinged Rack; Packaging: HRS, 10 x 96
Pack of 960 for $63.96
Pack of 960 for $44.77 Online Instant Savings (30%)
Case of 5 PK for $287.00
Case of 5 PK for $200.90 Online Instant Savings (30%)
Check Availability
Add
to Cart
(0) in My Cart
02-707-423 Available on GSA/VA Contract for Federal Government customers only.  Yellow; Nonsterile; Hinged Rack; Packaging: RD, 10 x 96
Pack of 960 for $48.79
Pack of 960 for $34.15 Online Instant Savings (30%)
Case of 5 PK for $208.07
Case of 5 PK for $145.65 Online Instant Savings (30%)
Check Availability
Add
to Cart
(0) in My Cart
Add Items to Cart
Description
  • All racked sterile tips are shrink wrapped and certified free of RNase, DNase, ATP, pyrogen, bioburden, PCR inhibitors, endotoxin, and DNA
  • All nonsterile racked and bulk nonfiltered tips are certified free of RNase, DNase, and DNA
SureOne Rack Features
  • Flush Fit Inserts sit flush with rack, allowing easy multichannel pipetting
  • Notched clasp props lid open for autoclave venting
  • Sure Stack lid rim prevents racks from sliding around when stacked and carted around a lab
  • Safe-Lock latch allows for one-handed open and closing ability and won't pop open when dropped
  • Sure-snap design securely holds insert in place, preventing it from being lifted out while pipetting
  • Reversible flip-lid can be either a hinged or lift-off lid; hinged format does not prevent use of a multichannel pipet
  • Recycle logo molded into all rack components
SureOne Insert Features
  • Inserts are easy to place onto and remove from rack; clip onto rack for a secure fit
  • Lot traceable — all inserts contain the Fisherbrand part number, tip volume, lot number
  • Indented insert holes inhibit tip movement, stabilizing them for multichannel work
  • Contact clip prevents unnecessary contact with the tips while clipping in inserts
  • Multichannel design; all inserts are 96-well format
  • Robust non-pliable design provides secure seals on every tip
SureOne Reload Packaging Features
  • Product information displayed on box
  • Dust flap prevents debris from contaminating the product
  • Finger knockouts allow easy access to slide out product
  • Canyon cut package allows for easy tip insert removal
  • Reload packaging design saves 58% bench space vs. racked packaging; reduces plastic waste by 52%
Specifications & Ordering Information:
Packaging options include bulk, rack, rack sterile, and reload.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your 9-digit account number, or become a registered user on fishersci.com by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service at 1-800-766-7000.