Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Print Email
Sciteck™ SVT™ Adulteration Creatinine Reagent
For use with automated systems for drugs of abuse adulteration testing
  • For in vitro diagnostic use to detect presence of creatinine in serum and urine
  • In an akaline medium, creatinine forms a yellow-orange colored complex with picric acid; color intensity is directly proportional to concentration of creatinine in urine and measured spectrophotmetrically at 505 nm
  • NaOH R1, picric acid R2, and calibrator solutions are ready for use
View Specifications
Images for Sciteck™ SVT™ Adulteration Creatinine Reagent
(6 item(s) in product Group)
  • Items
  • Specifications
Catalog No. Description Price per Unit Quantity & Availability Add Item(s)
23-424-4532
Sciteck Diagnostics
No.:134-30
Creatinine Reagent, R1 NaOH; 750mL
Each for $149.14
Check Availability
Add
to Cart
(0) in My Cart
23-424-4533
Sciteck Diagnostics
No.:135-30
Creatinine Reagent, R2 Picric Acid; 150mL
Each for $146.26
Check Availability
Add
to Cart
(0) in My Cart
23-424-4531
Sciteck Diagnostics
No.:139-30
Svt Creat R1/r2 450/300ml/pk
Pack of 2 for $374.59
Check Availability
Add
to Cart
(0) in My Cart
23-424-4534
Sciteck Diagnostics
No.:132-01
Creatinine Calibrator, 20mg/dL; 60mL
Each for $73.87
Check Availability
Add
to Cart
(0) in My Cart
23-424-4603
Sciteck Diagnostics
No.:136-01
Creatinine Calibrator, 100mg/dL; 60mL
Each for $71.03
Check Availability
Add
to Cart
(0) in My Cart
23-424-4535
Sciteck Diagnostics
No.:137-01
Creatinine Calibrator, 400mg/dL; 60mL
Each for $71.81
Check Availability
Add
to Cart
(0) in My Cart
Add Items to Cart
Description
  • For in vitro diagnostic use to detect presence of creatinine in serum and urine
  • In an akaline medium, creatinine forms a yellow-orange colored complex with picric acid; color intensity is directly proportional to concentration of creatinine in urine and measured spectrophotmetrically at 505 nm
  • NaOH R1, picric acid R2, and calibrator solutions are ready for use
Sciteck™ Specimen Validity Testing (SVT™)
  • In accordance with SAMHSA's FWDTP guidelines as specified by Executive Order 12564 and delineated in the Federal Register Vol. 53 FR 11970-11989 and Vol. 59 FR 29908-29931; and other federally mandated programs (DOT, DOE, NRC and FHWA) for employment drug screening
  • Sciteck evolved from Chimera Research, providing liquid reagents to laboratories for specimen validity testing in urine specimens submitted for DAU testing since 1989
  • Adulteration testing products to include extensive SVT adulteration and EIA™ Drugs of Abuse single and multi-assay(s) (4-in-1™) reagent line
  • SVT line consists of assays for pH, specific gravity, nitrite (non-corrosive), aldehydes, oxidants, chromate, creatinine, and halogens (bleach)
  • AdultaCheck™ for on-site adulteration products
Recommended Storage: Reagents and calibrators are stable until expiration date