Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Print Email
Sciteck™ SVT™ Adulteration pH Reagent
For in vitro diagnostic use to determine pH of a solution
  • Colorimetric: endpoint
  • Based on indicator principle which gives a broad range of color intensity covering entire urinary pH range
  • May be used on freshly voided urine specimen or urine collected under special conditions (e.g. first-morning specimens and post-prandial urine)
View Specifications
Images for Sciteck™ SVT™ Adulteration pH Reagent
(12 item(s) in product Group)
  • Items
  • Specifications
Catalog No. Description Price per Unit Quantity & Availability Add Item(s)
23-424-4514
Sciteck Diagnostics
No.:100
pH Reagent, pH Reagent R (1); 900mL
Each for $715.46
Check Availability
Add
to Cart
(0) in My Cart
23-424-4524
Sciteck Diagnostics
No.:115
pH Reagent, Blank Reagent for use on Olympus; 900mL
Each for $90.41
Check Availability
Add
to Cart
(0) in My Cart
23-424-4515
Sciteck Diagnostics
No.:107
pH Reagent, pH Buffers 4.5 and 9.0; 2 x 60mL
Pack of 2 for $321.78
Check Availability
Add
to Cart
(0) in My Cart
23-424-4521
Sciteck Diagnostics
No.:107L
pH Reagent, pH Buffers 4.5 and 9.0; 2 x 900mL
Pack of 2 for $629.18
Check Availability
Add
to Cart
(0) in My Cart
23-424-4516
Sciteck Diagnostics
No.:109
pH Reagent, pH Buffers 5.0 and 8.0; 2 x 60mL
Pack of 2 for $307.19
Check Availability
Add
to Cart
(0) in My Cart
23-424-4517
Sciteck Diagnostics
No.:107-4.5
pH Reagent, pH Buffer 4.5; 60mL
Each for $155.36
Check Availability
Add
to Cart
(0) in My Cart
23-424-4522
Sciteck Diagnostics
No.:107L4
pH Reagent, pH Buffer 4.5; 900mL
Each for $468.92
Check Availability
Add
to Cart
(0) in My Cart
23-424-4519
Sciteck Diagnostics
No.:109-5.0
pH Reagent, pH Buffer 5.0; 60mL
Each for $155.36
Check Availability
Add
to Cart
(0) in My Cart
23-424-4520
Sciteck Diagnostics
No.:109-7.0
pH Reagent, pH Buffer 7.0; 60mL
Each for $155.36
Check Availability
Add
to Cart
(0) in My Cart
23-424-4518
Sciteck Diagnostics
No.:107-9.0
pH Reagent, pH Buffer 9.0; 60mL
Each for $155.36
Check Availability
Add
to Cart
(0) in My Cart
23-424-4523
Sciteck Diagnostics
No.:107L9
pH Reagent, pH Buffer 9.0; 900mL
Each for $468.92
Check Availability
Add
to Cart
(0) in My Cart
23-424-4591
Sciteck Diagnostics
No.:114
pH meter calibrator; pH Buffer 12.0; 900mL
Each for $375.13
Check Availability
Add
to Cart
(0) in My Cart
Add Items to Cart
Description
  • Colorimetric: endpoint
  • Based on indicator principle which gives a broad range of color intensity covering entire urinary pH range
  • May be used on freshly voided urine specimen or urine collected under special conditions (e.g. first-morning specimens and post-prandial urine)
Sciteck™ Specimen Validity Testing (SVT™)
  • In accordance with SAMHSA's FWDTP guidelines as specified by Executive Order 12564 and delineated in the Federal Register Vol. 53 FR 11970-11989 and Vol. 59 FR 29908-29931; and other federally mandated programs (DOT, DOE, NRC and FHWA) for employment drug screening
  • Sciteck evolved from Chimera Research, providing liquid reagents to laboratories for specimen validity testing in urine specimens submitted for DAU testing since 1989
  • Adulteration testing products to include extensive SVT adulteration and EIA™ Drugs of Abuse single and multi-assay(s) (4-in-1™) reagent line
  • SVT line consists of assays for pH, specific gravity, nitrite (non-corrosive), aldehydes, oxidants, chromate, creatinine, and halogens (bleach)
  • AdultaCheck™ for on-site adulteration products
Recommended Storage: Store at 2-10°C; stable until expiration date
Disclaimer:
U.S. and Foreign Patents and Patents Pending: 6,537,823; 5,759,860.