Customer Service:  +1 800-766-7000Web Support:  +1 877-885-2081All Other Contact Info
Print Email
14-558-643
Sartorius™ BIOSTAT™ A plus Fermentor – Bioreactor System
The introductory Fermentor – Bioreactor System
BIOSTAT™ Aplus is a compact, autoclavable laboratory fermentor – bioreactor system suitable for a wide range of research, education and industrial applications. A number of easy-to-use, pre-configured versions are available for both microbial culture and cell culture which simplify the handling of the fermentor. The space saving control unit with its universal motor fits all available vessel sizes (1L/2L/5L)
Cell Culture package
  • Suitable for Mammalian, Insect and Plant Cell Culture
  • Working Volumes from 0.4 to 5L
  • Integrated 4-gas mixing system (Air, O2, N2, CO2)
  • Freely selectable gas composition to sparger or overlay (headspace)
  • 4 Flow Meters for individual flow rate setting
  • DO control via automatic valves for Air and O2
  • pH control via CO2 / Acid and Base
  • Special Impeller Design for low shear stress
Microbial package
  • Suitable for Bacteria, Yeast and Fungi cultivation
  • Working Volumes from 0.4 to 5L
  • Airflow gassing system with oxygen enrichment capability
  • Efficient agitation system for high cell density cultures
  • Automatic DO control for agitation, air and O2 flow or substrate addition
  • Anaerobic culture
View Specifications
Images for Sartorius™ BIOSTAT™ A plus Fermentor – Bioreactor System
  • 14-558-643
14-558-643
14-558-643
(6 item(s) in product Group)
  • Items
  • Specifications
Catalog No. Description Price per Unit Quantity & Availability Add Item(s)
14-558-643 Available on GSA/VA Contract for Federal Government customers only. 
Sartorius Stedim North America Inc.
No.:RAP-C01LFTSDM1
Cell Culture, 1L
14-558-613 Available on GSA/VA Contract for Federal Government customers only. 
Sartorius Stedim North America Inc.
No.:RAP-C02LFTSDM1
Cell Culture, 2L
14-558-614 Available on GSA/VA Contract for Federal Government customers only. 
Sartorius Stedim North America Inc.
No.:RAP-C05LFTSDM1
Cell Culture, 5L
14-558-615 Available on GSA/VA Contract for Federal Government customers only. 
Sartorius Stedim North America Inc.
No.:RAP-M01LOTRDM1
Microbial, 1L
14-558-616 Available on GSA/VA Contract for Federal Government customers only. 
Sartorius Stedim North America Inc.
No.:RAP-M02LOTRDM1
Microbial, 2L
14-558-617 Available on GSA/VA Contract for Federal Government customers only. 
Sartorius Stedim North America Inc.
No.:RAP-M05LOTRDM1
Microbial, 5L
Description
Cell Culture package
  • Suitable for Mammalian, Insect and Plant Cell Culture
  • Working Volumes from 0.4 to 5L
  • Integrated 4-gas mixing system (Air, O2, N2, CO2)
  • Freely selectable gas composition to sparger or overlay (headspace)
  • 4 Flow Meters for individual flow rate setting
  • DO control via automatic valves for Air and O2
  • pH control via CO2 / Acid and Base
  • Special Impeller Design for low shear stress
Microbial package
  • Suitable for Bacteria, Yeast and Fungi cultivation
  • Working Volumes from 0.4 to 5L
  • Airflow gassing system with oxygen enrichment capability
  • Efficient agitation system for high cell density cultures
  • Automatic DO control for agitation, air and O2 flow or substrate addition
  • Anaerobic culture
Bioreactor Operation
A powerful Notebook PC is included in every BIOSTAT™ Aplus. Thecolor graphical operation screen is self-explanatory and simple to operate. Up to 4 bioreactors | fermentors can be operated by one Notebook PC. The user can define specific names for each unit which makes identification and interaction between the processes very efficient.
Data Aquisition
BioPAT™ MFCS/DA (Data Acquisition), the world standard in bioprocess control, is included in every delivery. It offers a broad range of features including:
Batch-oriented data collection and storage
  • Evaluation of on-line and off-line measurements or calculated values
  • Data of finished or active batches can be exported to third party programs (e.g. MS Excel)